missing translation for 'onlineSavingsMsg'
Learn More
Trifluoromethanesulfonic acid, 99%, extra pure, Thermo Scientific Chemicals Product Code.: 10397440

Trifluoromethanesulfonic acid, 99%, extra pure, Thermo Scientific Chemicals

Product Code. 10397440
10 mL, Glass bottle
Click to view available options
Quantity:
1 mL
10 mL
50 mL
250 mL
1 L
Packaging:
Glass bottle
This item is not returnable. View return policy

Product Code. 10397440

Brand: Thermo Scientific Acros 169890100

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Trifluoromethanesulfonic acid, 99%, CHF3O3S, CAS Number-1493-13-6, hotf, methanesulfonic acid, trifluoro, tfmsa, perfluoromethanesulfonic acid, tfoh, trifluoromethanesulphonic acid, triflic acid, trifluoromethane sulfonic acid, trimsylate, trifluoromethylsulfonic acid, 250mL, 03,IV,34, 162.0 deg.C

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1493-13-6
Molecular Formula CHF3O3S
Molecular Weight (g/mol) 150.07
MDL Number MFCD00007514
InChI Key ITMCEJHCFYSIIV-UHFFFAOYSA-N
Synonym triflic acid, perfluoromethanesulfonic acid, trifluoromethanesulphonic acid, tfmsa, trifluoromethane sulfonic acid, tfoh, methanesulfonic acid, trifluoro, hotf, trifluoromethylsulfonic acid, trimsylateShow MoreShow Less
PubChem CID 62406
ChEBI CHEBI:48511
SMILES OS(=O)(=O)C(F)(F)F

Specifications

Melting Point -40.0°C
Color Colorless to Yellow
Density 1.6900g/mL
Boiling Point 162.0°C
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Linear Formula CF3SO3H
Refractive Index 1.3260 to 1.3300
Quantity 10 mL
Beilstein 03,IV,34
Fieser 04,532; 05,701; 06,617; 08,504; 09,531; 14,152; 15,41; 16,357; 17,24
Merck Index 15,9842
Specific Gravity 1.69
Solubility Information Solubility in water: soluble. Other solubilities: soluble in many polar organic solvents such as dim,sulfolane,dimethyl sulfoxide,dimethyl sulfone an
IUPAC Name trifluoromethanesulfonic acid
Viscosity 2.87 mPa.s (25°C)
Formula Weight 150.07
Percent Purity 99%
Grade Extra Pure
Physical Form Fuming Liquid
Chemical Name or Material Trifluoromethanesulfonic acid
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0465445 Certificate TypeCertificate of Analysis Date19/08/2024 Catalog Number169890011,169890010,169890100,169892500,169890500
Lot NumberA0455349 Certificate TypeCertificate of Analysis Date16/02/2024 Catalog Number169890011,169890010,169890100,169892500,169890500
Lot NumberA0447877 Certificate TypeCertificate of Analysis Date14/12/2023 Catalog Number169890011,169890010,169890100,169892500,169890500
Lot NumberA0397688 Certificate TypeCertificate of Analysis Date15/12/2022 Catalog Number169890011,169890010,169890100,169892500,169890500
Lot NumberA0440187 Certificate TypeCertificate of Analysis Date14/10/2022 Catalog Number169890011,169890010,169890100,169892500,169890500
compliance-icons compliance-icons
Product Identifier
  • Trifluoromethanesulfonic acid
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Substances and mixtures corrosive to metals Category 1
  • Skin corrosion/irritation Category 1 B
  • Specific target organ toxicity Category 3
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
  • H290-May be corrosive to metals.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Trifluoromethanesulfonic acid, 99%, extra pure, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.