missing translation for 'onlineSavingsMsg'
Learn More

Trifluoromethanesulfonic acid solution, Honeywell™

Catalog Number 15604930
Click to view available options
Quantity:
1 L
This item is not returnable. View return policy
This item is not returnable. View return policy

Volumetric, 0.1 M CF3SO3H in acetic acid (0.1N), for titration in non-aqueous medium

Honeywell´s high-quality titration products cover the complete titration workflow and include:

  • Solutions for volumetric titration (acids, bases, or salts)
  • Buffers available as concentrates or ready-to-use

 

  • Reagents for complexometry:
    • Aminopolycarboxylic acids (EDTA/NTA analogs)
    • Masking agents
  • Indicators for volumetric and complexometric titrations

 


 

TRUSTED_SUSTAINABILITY
CAS 1493-13-6
Flash Point 38°C (100.4°F)
Quantity 1 L
Molecular Formula CHF3O3S
Linear Formula CF3SO3H
MDL Number MFCD00007514
UN Number UN2789
Beilstein 3693487
Synonym triflic acid, perfluoromethanesulfonic acid, trifluoromethanesulphonic acid, tfmsa, trifluoromethane sulfonic acid, tfoh, methanesulfonic acid, trifluoro, hotf, trifluoromethylsulfonic acid, trimsylate
InChI Key ITMCEJHCFYSIIV-UHFFFAOYSA-N
SMILES OS(=O)(=O)C(F)(F)F
IUPAC Name trifluoromethanesulfonic acid
Molecular Weight (g/mol) 150.07
PubChem CID 62406
ChEBI CHEBI:48511
Formula Weight 150.08g/mol
Chemical Name or Material Trifluoromethanesulfonic acid solution
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Trifluoromethanesulfonic acid solution
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 1 A
Hazard Statement
  • H226-Flammable liquid and vapour.
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P243-Take precautionary measures against static discharge.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P284-¢In case of inadequate ventilation! wear respiratory protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
missing translation for 'delimitedSupplementalStatement'
  • MIXTURE LIST-Contains : acetic acid
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'