Learn More
Trifluoromethanesulfonic anhydride, 98%, Thermo Scientific Chemicals
Alfa Aesar Trifluoromethanesulfonic anhydride, 98%, Quantity: 250g, Boiling Point: 81 deg.C to 83 deg.C, Melting Point: -80 deg.C, Molecular Weight: 282.127, Percent Purity: 98%, Beilstein: 1813600, CAS: 358-23-6, ChEBI: CHEBI:48509, Density: 1.72
£57.90 - £713.00
Chemical Identifiers
CAS | 358-23-6 |
---|---|
Molecular Formula | C2F6O5S2 |
Molecular Weight (g/mol) | 282.127 |
MDL Number | MFCD00000408 |
InChI Key | WJKHJLXJJJATHN-UHFFFAOYSA-N |
Synonym | trifluoromethanesulfonic anhydride, triflic anhydride, trifluoromethanesulphonic anhydride, trifluoromethanesulfonic acid anhydride, methanesulfonic acid, trifluoro-, anhydride, trifluoromethanesulphonic acid anhydride, unii-8w034lhg1u, tf2o, trifluoromethane sulfonyl trifluoromethanesulfonate, trifluoromethane sulfonic anhydride |
PubChem CID | 67749 |
ChEBI | CHEBI:48509 |
IUPAC Name | trifluoromethylsulfonyl trifluoromethanesulfonate |
SMILES | C(F)(F)(F)S(=O)(=O)OS(=O)(=O)C(F)(F)F |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11443910
|
Thermo Scientific Alfa Aesar
A11767.09 |
10 g |
£57.90
10g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11439333
|
Thermo Scientific Alfa Aesar
A11767.14 |
25 g |
£116.00
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11453910
|
Thermo Scientific Alfa Aesar
A11767.18 |
50 g |
£193.00
50g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11449333
|
Thermo Scientific Alfa Aesar
A11767.22 |
100 g |
£327.00
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11463910
|
Thermo Scientific Alfa Aesar
A11767.30 |
250 g |
£713.00
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Trifluoromethanesulfonic anhydride is used to convert phenols and imine into triflic ester and NTf group. It is a strong electrophile used for the introduction of triflyl group in chemical synthesis. It serves as a reagent in the preparation of alkyl and vinyl triflates, and for the stereoselective synthesis of mannosazide methyl uronate donors. It acts as a catalyst for glycosylation with anomeric hydroxy sugars to prepare polysaccharides.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsTrifluoromethanesulfonic anhydride is used to convert phenols and imine into triflic ester and NTf group. It is a strong electrophile used for the introduction of triflyl group in chemical synthesis. It serves as a reagent in the preparation of alkyl and vinyl triflates, and for the stereoselective synthesis of mannosazide methyl uronate donors. It acts as a catalyst for glycosylation with anomeric hydroxy sugars to prepare polysaccharides.
Solubility
Miscible with dichloromethane. Immiscible with hydrocarbons.
Notes
Moisture sensitive. Incompatible with water, strong bases, strong oxidizing agents, acids and alcohols.
Chemical Identifiers
358-23-6 | |
282.127 | |
WJKHJLXJJJATHN-UHFFFAOYSA-N | |
67749 | |
trifluoromethylsulfonyl trifluoromethanesulfonate |
C2F6O5S2 | |
MFCD00000408 | |
trifluoromethanesulfonic anhydride, triflic anhydride, trifluoromethanesulphonic anhydride, trifluoromethanesulfonic acid anhydride, methanesulfonic acid, trifluoro-, anhydride, trifluoromethanesulphonic acid anhydride, unii-8w034lhg1u, tf2o, trifluoromethane sulfonyl trifluoromethanesulfonate, trifluoromethane sulfonic anhydride | |
CHEBI:48509 | |
C(F)(F)(F)S(=O)(=O)OS(=O)(=O)C(F)(F)F |
Specifications
358-23-6 | |
1.72 | |
C2F6O5S2 | |
1.321 | |
UN3265 | |
Moisture sensitive | |
Miscible with dichloromethane. Immiscible with hydrocarbons. | |
C(F)(F)(F)S(=O)(=O)OS(=O)(=O)C(F)(F)F | |
282.127 | |
CHEBI:48509 | |
98% |
-80°C | |
81°C to 83°C | |
(CF3SO2)2O | |
MFCD00000408 | |
1813600 | |
trifluoromethanesulfonic anhydride, triflic anhydride, trifluoromethanesulphonic anhydride, trifluoromethanesulfonic acid anhydride, methanesulfonic acid, trifluoro-, anhydride, trifluoromethanesulphonic acid anhydride, unii-8w034lhg1u, tf2o, trifluoromethane sulfonyl trifluoromethanesulfonate, trifluoromethane sulfonic anhydride | |
WJKHJLXJJJATHN-UHFFFAOYSA-N | |
trifluoromethylsulfonyl trifluoromethanesulfonate | |
67749 | |
282.13 | |
Trifluoromethanesulfonic anhydride |
Safety and Handling
GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
P234-P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P390-P501c
H290-H302-H314-H335
missing translation for 'dotInformation' : Hazard Class: 8; Packaging Group: I
missing translation for 'einecsNumber' : 206-616-8
missing translation for 'rtecsNumber' : PB2772000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
- Trifluoromethanesulfonic anhydride
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 1
- Substances and mixtures corrosive to metals Category 1
- Skin corrosion/irritation Category 1 A
Hazard Statement
- H290-May be corrosive to metals.
- H302-Harmful if swallowed.
- H314-Causes severe skin burns and eye damage.
- H318-Causes serious eye damage.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
- MIXTURE LIST-Contains : trifluoro-Methanesulfonic acid anhydride
RUO – Research Use Only