Learn More
Invitrogen™ Triiodothyronine (T3) Competitive ELISA Kit
Competitive ELISA
£548.00 - £3590.00
Specifications
Assay Sensitivity | 37.4 pg/mL |
---|---|
Description | Triiodothyronine (T3) Competitive ELISA Kit |
Conjugate | HRP |
Detection Method | Fluorescence |
Product Type | ELISA Kit |
Product Code | Brand | Sufficient For | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Sufficient For | Price | Quantity & Availability | |||||
15835654
|
Invitrogen™
EIAT3C |
96 assays |
£548.00
96 tests |
Estimated Shipment: 12-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
17845017
|
Invitrogen™
EIAT3CX10 |
10 x 96 Assays |
£3590.00
Pack of 10 |
Estimated Shipment: 28-08-2024
Log in to see stock. |
|||||
Description
The Triiodothyronine (T3) ELISA quantitates T3 in serum,plasma,urine,dried fecal extracts,or cell culture medium.
Principle of the method
The Triiodothyronine (T3) Competitive ELISA research-use-only kit is designed to quantitatively measure T3 present in extracted serum and plasma,urine,extracted dried fecal samples,and tissue culture media samples. This kit measures total T3 in extracted serum and plasma and in extracted fecal samples. A T3 stock solution is provided to generate standard curves for the assay and all samples should be read off the standard curve. Standards or diluted samples are pipetted into a clear microtiter plate coated with an antibody to capture sheep antibodies. A T3-peroxidase conjugate is added to the standards and samples in the wells. The binding reaction is initiated by the addition of a sheep antibody to T3 to each well. After a two-hour incubation the plate is washed and substrate is added. The substrate reacts with the bound T3-peroxidase conjugate. After a short incubation,the reaction is stopped and the intensity of the generated color is detected in a microtiter plate reader capable of measuring at 450 nm.
Rigorous validation
Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity,specificity,precision,and lot-to-lot consistency. See manual for more information on validation.
Specifications
37.4 pg/mL | |
HRP | |
ELISA Kit | |
Human | |
Colorimetric Microplate Reader | |
6.3% | |
RUO | |
-20°C | |
1 hr. 20 min. |
Triiodothyronine (T3) Competitive ELISA Kit | |
Fluorescence | |
Fecal Extract, Plasma, Serum, Supernatant, Urine | |
ELISA, Protein Assays, Aktivity Assays, Protein Biology, Reproductive Biology, Inflammation, Oxidative Stress, Metabolism, Cell Signaling, Epigenetics, Hematology, DNA Damage, Cancer Biology, Immunology, Neuroscience, Neurobiology, Steroid Hormone Detecti | |
13.4% | |
HRP | |
Metabolism | |
Chemical | |
2 hr. 30 min. |