Learn More
Triisobutylaluminium, 1.1M solution in toluene, AcroSeal™, Thermo Scientific Chemicals
£51.50 - £312.00
Chemical Identifiers
CAS | 100-99-2, 108-88-3 |
---|---|
Molecular Formula | C12H27Al |
Molecular Weight (g/mol) | 198.33 |
MDL Number | MFCD00008929 |
InChI Key | MCULRUJILOGHCJ-UHFFFAOYSA-N |
Synonym | triisobutylaluminum, triisobutylaluminium, aluminum, tris 2-methylpropyl, tibal, triisobutylalane, triisobutyl aluminum, aluminum, triisobutyl, tris isobutyl alane, tris isobutyl aluminum, unii-09p2thv2x4 |
PubChem CID | 16682931 |
IUPAC Name | tris(2-methylpropyl)alumane |
SMILES | CC(C)C[Al](CC(C)C)CC(C)C |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10178692
|
Thermo Scientific Acros
185421000 |
100 mL | AcroSeal™ Glass bottle |
£51.50
100mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10301753
|
Thermo Scientific Acros
185428000 |
800 mL | AcroSeal™ Glass Bottle |
£312.00
800mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Lewis acidChemical Identifiers
100-99-2, 108-88-3 | |
198.33 | |
MCULRUJILOGHCJ-UHFFFAOYSA-N | |
16682931 | |
CC(C)C[Al](CC(C)C)CC(C)C |
C12H27Al | |
MFCD00008929 | |
triisobutylaluminum, triisobutylaluminium, aluminum, tris 2-methylpropyl, tibal, triisobutylalane, triisobutyl aluminum, aluminum, triisobutyl, tris isobutyl alane, tris isobutyl aluminum, unii-09p2thv2x4 | |
tris(2-methylpropyl)alumane |
Specifications
Colorless to Yellow | |
C12H27Al | |
MFCD00008929 | |
0.8400g/mL | |
triisobutylaluminum, triisobutylaluminium, aluminum, tris 2-methylpropyl, tibal, triisobutylalane, triisobutyl aluminum, aluminum, triisobutyl, tris isobutyl alane, tris isobutyl aluminum, unii-09p2thv2x4 | |
MCULRUJILOGHCJ-UHFFFAOYSA-N | |
tris(2-methylpropyl)alumane | |
16682931 | |
24.5 to 25.5% (on Al basis) | |
4°C | |
Triisobutylaluminium, 1.1M Solution in Toluene |
100-99-2, 108-88-3 | |
[(CH3)2CHCH2]3Al | |
09,486 | |
10, 316 | |
Solubility in water: reacts. Other solubilities: miscible with aromatic and aliphatic hydrocarbons | |
CC(C)C[Al](CC(C)C)CC(C)C | |
198.33 | |
198.33 | |
0.84 | |
Solution |
Safety and Handling
GHS H Statement
May be fatal if swallowed and enters airways. Causes severe skin burns and eye damage. May cause drowsiness or dizziness. May cause damage to organs through prolonged or repeated exposure. Suspected of damaging the unborn child. Highly flammable liquid and vapor. In contact with water releases flammable gases which may ignite spontaneously. Reacts violently with water.
GHS P Statement
IF SWALLOWED: rinse mouth. Do NOT induce vomiting. Wear eye protection/face protection. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Immediately call a POISON CENTER or doctor/physician. Use personal protective equipment as required. IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing. Do not breathe dust/fume/gas/mist/vapors/spray. IF SWALLOWED: Immediately call a POISON CENTRE or doctor/physician. Do NOT induce vomiting. Store in a dry place. Store in a closed container.
Danger
- Triisobutylaluminium, solution in toluene
Signal Word
- Danger
Hazard Category
- Aspiration toxicity Category 1
- Serious eye damage/eye irritation Category 1
- Flammable liquid Category 2
- Reproductive toxicity Category 2
- Skin corrosion/irritation Category 1 B
- Specific target organ toxicity after repeated exposure Category 2
- Specific target organ toxicity Category 3
- Substance which\, in contact with water\, emits flammable gases Category 1
Hazard Statement
- H304-May be fatal if swallowed and enters airways.
- H318-Causes serious eye damage.
- H314-Causes severe skin burns and eye damage.
- H361d-Suspected of damaging the unborn child.
- H373-May cause damage to organs.
- H336-May cause drowsiness or dizziness.
- H260-In contact with water releases flammable gases which may ignite spontaneously.
- H225-Highly flammable liquid and vapour.
- EUH014-Reacts violently with water.
Precautionary Statement
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- P331-Do NOT induce vomiting.
- P402+P404-Store in a dry place. Store in a closed container.
RUO – Research Use Only