missing translation for 'onlineSavingsMsg'
Learn More

Triisopropanolamine, 98%, Thermo Scientific Chemicals

£10.45 - £140.70

Chemical Identifiers

CAS 122-20-3
Molecular Formula C9H21NO3
Molecular Weight (g/mol) 191.27
MDL Number MFCD00004533
InChI Key SLINHMUFWFWBMU-UHFFFAOYSA-N
Synonym triisopropanolamine, tipa, tri-2-propanolamine, tri-iso-propanolamine, tris 2-propanol amine, 2-propanol, 1,1',1-nitrilotris, tris 2-hydroxypropyl amine, 1,1',1-nitrilotripropan-2-ol, tris 2-hydroxy-1-propyl amine, caswell no. 891
PubChem CID 24730
IUPAC Name 1-[bis(2-hydroxypropyl)amino]propan-2-ol
SMILES CC(CN(CC(C)O)CC(C)O)O
View More Specs

Products 4
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10479930
View Documents
Thermo Scientific Acros
245760050
5 g Glass bottle This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
10279453
View Documents Promotion Details
Thermo Scientific Acros
245761000
100 g Glass bottle
£17.79
100g
Please to purchase this item. Need a web account? Register with us today!
10388030
View Documents
Thermo Scientific Acros
245760010
1 kg Glass bottle
£25.33
1kg
Please to purchase this item. Need a web account? Register with us today!
13488488
View Documents
Thermo Scientific Acros
245760100
10 kg Plastic Can
£140.70
10kg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

122-20-3
191.27
SLINHMUFWFWBMU-UHFFFAOYSA-N
24730
CC(CN(CC(C)O)CC(C)O)O
C9H21NO3
MFCD00004533
triisopropanolamine, tipa, tri-2-propanolamine, tri-iso-propanolamine, tris 2-propanol amine, 2-propanol, 1,1',1-nitrilotris, tris 2-hydroxypropyl amine, 1,1',1-nitrilotripropan-2-ol, tris 2-hydroxy-1-propyl amine, caswell no. 891
1-[bis(2-hydroxypropyl)amino]propan-2-ol

Specifications

122-20-3
White to Yellow
160°C
98%
N(CH2CH(OH)CH3)3
04,762
triisopropanolamine, tipa, tri-2-propanolamine, tri-iso-propanolamine, tris 2-propanol amine, 2-propanol, 1,1',1-nitrilotris, tris 2-hydroxypropyl amine, 1,1',1-nitrilotripropan-2-ol, tris 2-hydroxy-1-propyl amine, caswell no. 891
SLINHMUFWFWBMU-UHFFFAOYSA-N
1-[bis(2-hydroxypropyl)amino]propan-2-ol
24730
191.27
Crystalline Low Melting Solid
48.0°C to 52.0°C
190.0°C (23.0 mmHg)
Authentic
C9H21NO3
MFCD00004533
17,371; 12,445; 13,46
Solubility in water: soluble. Other solubilities: soluble in ethanol, diethyl ether
CC(CN(CC(C)O)CC(C)O)O
191.27
138 mPa.s (60°C)
98%
Triisopropanolamine
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes serious eye irritation.
Harmful to aquatic life with long lasting effects.

GHS P Statement
Avoid release to the environment.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.

GHS Signal Word: Warning

missing translation for 'einecsNumber' : 204-528-4

Product Identifier
  • Triisopropanolamine

Signal Word
  • Warning

Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Serious eye damage/eye irritation Category 2

Hazard Statement
  • H412-Harmful to aquatic life with long lasting effects.
  • H319-Causes serious eye irritation.

Precautionary Statement
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only