missing translation for 'onlineSavingsMsg'
Learn More
Triisopropyl borate, 98+%, AcroSeal™, Thermo Scientific Chemicals Product Code.: 10070764

Triisopropyl borate, 98+%, AcroSeal™, Thermo Scientific Chemicals

Product Code. 10070764 Shop All Thermo Scientific Products
100 mL, AcroSeal™ Glass Bottle
Click to view available options
Quantity:
100 mL
800 mL
Packaging:
AcroSeal™ Glass Bottle
This item is not returnable. View return policy
This item is packed in AcroSeal™, the industry-leading packaging for air and moisture sensitive reagents.
Find out more about AcroSeal™ packaging.
 

Product Code. 10070764

Brand: Thermo Scientific 427141000

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
This item is packed in AcroSeal™, the industry-leading packaging for air and moisture sensitive reagents.
Find out more about AcroSeal™ packaging.
 

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5419-55-6
Molecular Formula C9H21BO3
Molecular Weight (g/mol) 188.07
MDL Number MFCD00008872
InChI Key NHDIQVFFNDKAQU-UHFFFAOYSA-N
Synonym triisopropyl borate, isopropyl borate, triisopropoxyborane, triisopropoxyboron, boron isopropoxide, triisopropyl orthoborate, boric acid triisopropyl ester, triisopropoxy borane, boron triisopropoxide, boric acid h3bo3 , tris 1-methylethyl ester
PubChem CID 21531
IUPAC Name tripropan-2-yl borate
SMILES B(OC(C)C)(OC(C)C)OC(C)C

Specifications

Melting Point -59°C
Color Colorless
Density 0.8140g/mL
Boiling Point 140°C
Flash Point 17°C
Infrared Spectrum Authentic
Assay Percent Range 98% min. (GC)
Packaging AcroSeal™ Glass Bottle
Linear Formula [(CH3)2CHO]3B
Refractive Index 1.3755 to 1.3775
Quantity 100 mL
Specific Gravity 0.814
Solubility Information Solubility in water: decomposes. Other solubilities: soluble in alcohol,ether and benzene
Formula Weight 188.07
Percent Purity 98+%
Physical Form Liquid
Chemical Name or Material Triisopropyl borate
Show More Show Less
compliance-icons
Product Identifier
  • Triisopropyl borate
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
Hazard Statement
  • H225-Highly flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P240-Ground/bond container and receiving equipment.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Triisopropyl borate, 98+%, AcroSeal™, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.