Learn More
Triisopropyl borate, 98+%, Thermo Scientific Chemicals
£30.30 - £145.00
Chemical Identifiers
CAS | 5419-55-6 |
---|---|
Molecular Formula | C9H21BO3 |
Molecular Weight (g/mol) | 188.07 |
MDL Number | MFCD00008872 |
InChI Key | NHDIQVFFNDKAQU-UHFFFAOYSA-N |
Synonym | triisopropyl borate, isopropyl borate, triisopropoxyborane, triisopropoxyboron, boron isopropoxide, triisopropyl orthoborate, boric acid triisopropyl ester, triisopropoxy borane, boron triisopropoxide, boric acid h3bo3 , tris 1-methylethyl ester |
PubChem CID | 21531 |
IUPAC Name | tripropan-2-yl borate |
SMILES | B(OC(C)C)(OC(C)C)OC(C)C |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10236630
|
Thermo Scientific Acros
188740800 |
80 g | Glass bottle |
£30.30
80g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10713524
|
Thermo Scientific Acros
188747500 |
750 g | Glass bottle |
£145.00
750g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
5419-55-6 | |
188.07 | |
NHDIQVFFNDKAQU-UHFFFAOYSA-N | |
21531 | |
B(OC(C)C)(OC(C)C)OC(C)C |
C9H21BO3 | |
MFCD00008872 | |
triisopropyl borate, isopropyl borate, triisopropoxyborane, triisopropoxyboron, boron isopropoxide, triisopropyl orthoborate, boric acid triisopropyl ester, triisopropoxy borane, boron triisopropoxide, boric acid h3bo3 , tris 1-methylethyl ester | |
tripropan-2-yl borate |
Specifications
5419-55-6 | |
Colorless | |
140°C | |
Authentic | |
C9H21BO3 | |
1.3755 to 1.3775 | |
01, 363 | |
triisopropyl borate, isopropyl borate, triisopropoxyborane, triisopropoxyboron, boron isopropoxide, triisopropyl orthoborate, boric acid triisopropyl ester, triisopropoxy borane, boron triisopropoxide, boric acid h3bo3 , tris 1-methylethyl ester | |
NHDIQVFFNDKAQU-UHFFFAOYSA-N | |
tripropan-2-yl borate | |
21531 | |
98+% | |
Triisopropyl borate |
-59°C | |
0.8140g/mL | |
17°C | |
98% min. (GC) | |
[(CH3)2CHO]3B | |
MFCD00008872 | |
0.814 | |
Solubility in water: decomposes. Other solubilities: soluble in alcohol,ether and benzene | |
B(OC(C)C)(OC(C)C)OC(C)C | |
188.07 | |
188.07 | |
Liquid |
Safety and Handling
GHS H Statement:
Highly flammable liquid and vapor.
GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 226-529-9
RUO – Research Use Only