missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Triisopropylphosphine, 90+%, Thermo Scientific Chemicals
Click to view available options
Quantity:
0.5 g
2 g
10 g
Description
Triisopropylphosphine is used as a ligand in organometallic chemistry. It is an important raw material and intermediate used in organic Synthesis, pharmaceuticals and agrochemicals. It is also used to produce Chlor(triisopropyl)phosphonium-dimesylamid at temperature of 40°C.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
Density | 0.839g/mL |
Boiling Point | 176°C to 178°C |
Flash Point | −17° (1°F) |
Odor | Pungent |
Refractive Index | 1.4670 |
Quantity | 2 g |
UN Number | UN2845 |
Sensitivity | Air Sensitive |
Formula Weight | 160.24 |
Percent Purity | ≥90% |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'