missing translation for 'onlineSavingsMsg'
Learn More

Triisopropylphosphine, 90+%, Thermo Scientific Chemicals

Product Code. 15431337
Click to view available options
Quantity:
0.5 g
2 g
10 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Triisopropylphosphine is used as a ligand in organometallic chemistry. It is an important raw material and intermediate used in organic Synthesis, pharmaceuticals and agrochemicals. It is also used to produce Chlor(triisopropyl)phosphonium-dimesylamid at temperature of 40°C.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6476-36-4
Molecular Formula C9H21P
Molecular Weight (g/mol) 160.241
MDL Number MFCD00015029
InChI Key IGNTWNVBGLNYDV-UHFFFAOYSA-N
Synonym triisopropylphosphine, tri-i-propylphosphine, phosphine, triisopropyl, phosphine, tris 1-methylethyl, tri-isopropylphosphine, triisopropylphosphane, trisisopropylphosphine, triisopropyl phosphine, acmc-20ajdu, pubchem6458
PubChem CID 80969
IUPAC Name tri(propan-2-yl)phosphane
SMILES CC(C)P(C(C)C)C(C)C

Specifications

Density 0.839 g/mL
Boiling Point 176°C to 178°C
Flash Point -17°C (1°F)
Odor Pungent
Refractive Index 1.467
Quantity 10 g
UN Number UN2845
Sensitivity Air Sensitive
Solubility Information Soluble in alkanes.
Formula Weight 160.24
Percent Purity ≥90%
Physical Form Liquid
Chemical Name or Material Triisopropylphosphine
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'