Learn More
(Triisopropylsilyl)acetylene, 97%, Thermo Scientific Chemicals
£59.60 - £192.96
Chemical Identifiers
CAS | 89343-06-6 |
---|---|
Molecular Formula | C11H22Si |
Molecular Weight (g/mol) | 182.38 |
MDL Number | MFCD00075452 |
InChI Key | KZGWPHUWNWRTEP-UHFFFAOYSA-N |
Synonym | triisopropylsilyl acetylene, ethynyltriisopropylsilane, triisopropylsilylacetylene, ethynyltris propan-2-yl silane, ethynyl-tri propan-2-yl silane, silane, ethynyltris 1-methylethyl, triisopropylsilylacetylen, acmc-209qzk, ethynyltriisopropyl-silane |
PubChem CID | 2734682 |
IUPAC Name | ethynyl-tri(propan-2-yl)silane |
SMILES | CC(C)[Si](C#C)(C(C)C)C(C)C |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10571702
|
Thermo Scientific Acros
368740050 |
5 g | Glass bottle |
£59.60
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10018981
|
Thermo Scientific Acros
368740250 |
25 g | Glass bottle |
£192.96
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
89343-06-6 | |
182.38 | |
KZGWPHUWNWRTEP-UHFFFAOYSA-N | |
2734682 | |
CC(C)[Si](C#C)(C(C)C)C(C)C |
C11H22Si | |
MFCD00075452 | |
triisopropylsilyl acetylene, ethynyltriisopropylsilane, triisopropylsilylacetylene, ethynyltris propan-2-yl silane, ethynyl-tri propan-2-yl silane, silane, ethynyltris 1-methylethyl, triisopropylsilylacetylen, acmc-209qzk, ethynyltriisopropyl-silane | |
ethynyl-tri(propan-2-yl)silane |
Specifications
89343-06-6 | |
100.0 | |
0.8130g/mL | |
56°C | |
96% min. (GC) | |
[(CH3)2CH]3SiC∽CH | |
MFCD00075452 | |
triisopropylsilyl acetylene, ethynyltriisopropylsilane, triisopropylsilylacetylene, ethynyltris propan-2-yl silane, ethynyl-tri propan-2-yl silane, silane, ethynyltris 1-methylethyl, triisopropylsilylacetylen, acmc-209qzk, ethynyltriisopropyl-silane | |
CC(C)[Si](C#C)(C(C)C)C(C)C | |
182.38 | |
182.38 | |
Liquid |
96.0 | |
Colorless | |
50°C to 52°C (6.0 mmHg) | |
Authentic | |
C11H22Si | |
1.4515 to 1.4535 | |
0.813 | |
KZGWPHUWNWRTEP-UHFFFAOYSA-N | |
ethynyl-tri(propan-2-yl)silane | |
2734682 | |
97% | |
(Triisopropylsilyl)acetylene, 97% |
Safety and Handling
GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
Flammable liquid and vapor.
GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Warning
- (Triisopropylsilyl)acetylene
Signal Word
- Warning
Hazard Category
- Flammable liquid Category 3
- Skin corrosion/irritation Category 2
- Serious eye damage/eye irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H226-Flammable liquid and vapour.
- H319-Causes serious eye irritation.
- H315-Causes skin irritation.
- H335-May cause respiratory irritation.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
RUO – Research Use Only