Learn More
TRIM45 Mouse anti-Human, Unconjugated, Clone: 3A7H11, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 66896-1-IG-150UL
This item is not returnable.
View return policy
Description
The TRIM45 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human TRIM45. This antibody recognizes human antigen. The TRIM45 antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
TRIM45 | |
Monoclonal | |
1000 μg/mL | |
Mouse | |
Primary | |
Human |
Western Blot, ELISA | |
3A7H11 | |
Unconjugated | |
150 μL | |
80263 | |
IgG1 |