missing translation for 'onlineSavingsMsg'
Learn More
Trimethoxyboroxine, Thermo Scientific™ Product Code.: 10648824

Trimethoxyboroxine, Thermo Scientific™

Product Code. 10648824
250 g, Glass bottle
Click to view available options
Quantity:
25 g
100 g
250 g
Packaging:
Glass bottle
This item is not returnable. View return policy

Product Code. 10648824

Brand: Thermo Scientific Acros 139962500

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 102-24-9
Molecular Formula C3H9B3O6
Molecular Weight (g/mol) 173.53
MDL Number MFCD00005946
InChI Key ZFMOJHVRFMOIGF-UHFFFAOYSA-N
Synonym trimethoxyboroxine, trimethoxyboroxin, boroxin, trimethoxy, trimethoxycyclotriboroxane, 2,4,6-trimethoxyboroxin, triboron trimethyl hexaoxide, boroxin, 2,4,6-trimethoxy, boric acid h3b3o6 , trimethyl ester, trimethoxy-boroxi, trimethoxyboroxoleShow MoreShow Less
PubChem CID 66880
IUPAC Name 2,4,6-trimethoxy-1,3,5,2,4,6-trioxatriborinane
SMILES COB1OB(OC)OB(OC)O1

Specifications

Melting Point 10°C to 11°C
Color Colorless
Density 1.2160g/mL
Flash Point 14°C
Packaging Glass bottle
Refractive Index 1.3996
Quantity 250 g
Specific Gravity 1.216
Solubility Information Solubility in water: reacts
Viscosity 21.7 mm2/s (20°C)
Formula Weight 173.53
Percent Purity 18 to 19.5% (as B)
Physical Form Liquid
Chemical Name or Material Trimethoxyboroxine
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

1 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0236244 Certificate TypeCertificate of Analysis Date12/08/2022 Catalog Number139962500,139960250,139961000

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Trimethoxyboroxine, Thermo Scientific™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.