missing translation for 'onlineSavingsMsg'
Learn More

Trimethoxysilane, 95%

Product Code. 10014293 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
5g
This item is not returnable. View return policy
Sea freight only: For EU stock allow up to 15 days delivery.

Product Code. 10014293

Brand: Thermo Scientific Acros 392171000

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Sea freight only: For EU stock allow up to 15 days delivery.

CAS: 2487-90-3 | C3H10O3Si | 122.2 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Reduction reagent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2487-90-3
Molecular Formula C3H10O3Si
Molecular Weight (g/mol) 122.2
MDL Number MFCD00008340
InChI Key PZJJKWKADRNWSW-UHFFFAOYSA-N
Synonym trimethoxysilane, silane, trimethoxy, trimethoxy silane, unii-v1j39xpf91, c3h10o3si, trimethoxysilyl, silanem trimethoxy, trimethoxysilyl radical, 4-01-00-01266 beilstein handbook reference
PubChem CID 6327428
IUPAC Name trimethoxysilicon
SMILES CO[Si](OC)OC

Specifications

Melting Point -115°C
Color Colorless
Density 0.9600g/mL
Boiling Point 86°C
Flash Point -9°C
Infrared Spectrum Authentic
Assay Percent Range 94% min. (GC)
Packaging Glass bottle
Linear Formula HSi(OCH3)3
Refractive Index 1.3570 to 1.3600 (20°C, 589nm)
Quantity 100 g
Specific Gravity 0.96
Solubility Information Solubility in water: reacts
Formula Weight 122.2
Percent Purity 95%
Physical Form Liquid
Chemical Name or Material Trimethoxysilane
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.