Learn More
Trimethylacetyl chloride, 99%, Thermo Scientific Chemicals
£16.20 - £72.70
Chemical Identifiers
CAS | 3282-30-2 |
---|---|
Molecular Formula | C5H9ClO |
Molecular Weight (g/mol) | 120.58 |
MDL Number | MFCD00000709 |
InChI Key | JVSFQJZRHXAUGT-UHFFFAOYSA-N |
Synonym | pivaloyl chloride, trimethylacetyl chloride, propanoyl chloride, 2,2-dimethyl, pivalyl chloride, pivaloylchloride, 2,2-dimethylpropionyl chloride, pivalic acid chloride, pivalolyl chloride, unii-jq82j0o21t, 2,2-dimethylpropionylchloride |
PubChem CID | 62493 |
IUPAC Name | 2,2-dimethylpropanoyl chloride |
SMILES | CC(C)(C)C(Cl)=O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10285090
|
Thermo Scientific Acros
140050050 |
5 g | Glass bottle |
£16.20
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10511771
|
Thermo Scientific Acros
140052500 |
250 g | Glass bottle |
£27.60
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10522744
|
Thermo Scientific Acros
140050010 |
1 kg | Glass bottle |
£72.70
1kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
3282-30-2 | |
120.58 | |
JVSFQJZRHXAUGT-UHFFFAOYSA-N | |
62493 | |
CC(C)(C)C(Cl)=O |
C5H9ClO | |
MFCD00000709 | |
pivaloyl chloride, trimethylacetyl chloride, propanoyl chloride, 2,2-dimethyl, pivalyl chloride, pivaloylchloride, 2,2-dimethylpropionyl chloride, pivalic acid chloride, pivalolyl chloride, unii-jq82j0o21t, 2,2-dimethylpropionylchloride | |
2,2-dimethylpropanoyl chloride |
Specifications
3282-30-2 | |
Colorless to Pink | |
105.0°C | |
Authentic | |
C5H9ClO | |
1.4110 to 1.4130 | |
02, 320 | |
0.98 | |
Solubility in water: hydrolysis., miscible with most common aprotic organic solvents | |
CC(C)(C)C(Cl)=O | |
120.58 | |
0.86 mPa.s (20°C) | |
99% | |
Trimethylacetyl chloride, 99% |
-56.0°C | |
0.9800g/mL | |
14°C | |
99% | |
(CH3)3CCOCl | |
MFCD00000709 | |
01,1229; 08,404; 11,489 | |
pivaloyl chloride, trimethylacetyl chloride, propanoyl chloride, 2,2-dimethyl, pivalyl chloride, pivaloylchloride, 2,2-dimethylpropionyl chloride, pivalic acid chloride, pivalolyl chloride, unii-jq82j0o21t, 2,2-dimethylpropionylchloride | |
JVSFQJZRHXAUGT-UHFFFAOYSA-N | |
2,2-dimethylpropanoyl chloride | |
62493 | |
120.58 | |
Liquid |
Safety and Handling
GHS H Statement
Harmful if swallowed.
Fatal if inhaled.
Causes severe skin burns and eye damage.
May be corrosive to metals.
Highly flammable liquid and vapor.
GHS P Statement
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if pre
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 221-921-6
missing translation for 'tsca' : TSCA
- Trimethylacetyl chloride
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 2
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 1
- Flammable liquid Category 2
- Substances and mixtures corrosive to metals Category 1
- Skin corrosion/irritation Category 1 B
Hazard Statement
- H318-Causes serious eye damage.
- H314-Causes severe skin burns and eye damage.
- H330-Fatal if inhaled.
- H302-Harmful if swallowed.
- H290-May be corrosive to metals.
- H225-Highly flammable liquid and vapour.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only