Learn More
Trimethylaluminium, 1.0M solution in heptane, AcroSeal™, Thermo Scientific Chemicals
£69.14 - £406.02
Chemical Identifiers
CAS | 75-24-1, 142-82-5 |
---|---|
Molecular Formula | C3H9Al |
Molecular Weight (g/mol) | 72.087 |
MDL Number | MFCD00008252 |
InChI Key | JLTRXTDYQLMHGR-UHFFFAOYSA-N |
Synonym | trimethylaluminum, trimethylaluminium, aluminum, trimethyl, trimethylalane, unii-av210lg46j, ch3 3al, aluminum trimethanide, aluminum trimethyl, trimethylaluminum solution, trimethylaluminum, elec. gr. |
PubChem CID | 16682925 |
IUPAC Name | trimethylalumane |
SMILES | C[Al](C)C |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10153502
|
Thermo Scientific Acros
189271000 |
100 mL | AcroSeal™ Glass bottle |
£69.14
100mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10211373
|
Thermo Scientific Acros
189278000 |
800 mL | AcroSeal™ Glass Bottle |
£406.02
800mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Descripción
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Lewis acidIdentificadores de productos químicos
75-24-1, 142-82-5 | |
72.087 | |
JLTRXTDYQLMHGR-UHFFFAOYSA-N | |
16682925 | |
C[Al](C)C |
C3H9Al | |
MFCD00008252 | |
trimethylaluminum, trimethylaluminium, aluminum, trimethyl, trimethylalane, unii-av210lg46j, ch3 3al, aluminum trimethanide, aluminum trimethyl, trimethylaluminum solution, trimethylaluminum, elec. gr. | |
trimethylalumane |
Especificaciones
Colorless | |
126.0°C | |
C3H9Al | |
MFCD00008252 | |
0.6880g/mL | |
trimethylaluminum, trimethylaluminium, aluminum, trimethyl, trimethylalane, unii-av210lg46j, ch3 3al, aluminum trimethanide, aluminum trimethyl, trimethylaluminum solution, trimethylaluminum, elec. gr. | |
JLTRXTDYQLMHGR-UHFFFAOYSA-N | |
trimethylalumane | |
16682925 | |
9.5 to 10.5 wt% (on Al basis) | |
0.90 mPa.s (30°C) | |
Liquid |
75-24-1, 142-82-5 | |
15.0°C | |
(CH3)3Al | |
08,506; 14,46; 17,372 | |
15, 9879 | |
Solubility in water: reacts. Other solubilities: miscible with saturated aliphatic and aromatic, hydrocarbons | |
C[Al](C)C | |
72.087 | |
72.09 | |
0.688 | |
−4°C | |
Trimethylaluminium, 1.0M Solution in Heptane |
Seguridad y manipulación
GHS H Statement:
May be fatal if swallowed and enters airways.
Very toxic to aquatic life with long lasting effects.
Causes severe skin burns and eye damage.
May cause drowsiness or dizziness.
In contact with water releases flammable gases which may ignite spontaneously.
GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTRE or doctor/physician.
Do NOT induce vomiting.
Avoid release to the environment.
Store in a dry place. Store in a closed container.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Danger
- Trimethylaluminium, 1.0M solution in heptane
Palabra de advertencia
- Danger
Clasificación
- LONG-TERM AQUATIC HAZARD Chronic 1
- ACUTE AQUATIC Acute 1
- Aspiration toxicity Category 1
- Serious eye damage/eye irritation Category 1
- Flammable liquid Category 2
- Skin corrosion/irritation Category 1 B
- Specific target organ toxicity Category 3
- Substance which\, in contact with water\, emits flammable gases Category 1
Indicaciones de peligro
- H304-May be fatal if swallowed and enters airways.
- H318-Causes serious eye damage.
- H400-Very toxic to aquatic life.
- H410-Very toxic to aquatic life with long lasting effects.
- H314-Causes severe skin burns and eye damage.
- H336-May cause drowsiness or dizziness.
- H260-In contact with water releases flammable gases which may ignite spontaneously.
- H225-Highly flammable liquid and vapour.
- EUH014-Reacts violently with water.
Consejos de prudencia
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P273-Avoid release to the environment.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- P331-Do NOT induce vomiting.
- P402+P404-Store in a dry place. Store in a closed container.
RUO – Research Use Only