Learn More
Trimethylamine, 1M solution in THF, AcroSeal™, Thermo Scientific Chemicals
£61.40 - £306.00
Specifications
Chemical Name or Material | Trimethylamine |
---|---|
Color | Colorless to Yellow |
Name Note | 1M solution in THF |
CAS | 75-50-3, 109-99-9 |
Molecular Formula | C3H9N |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10601464
|
Thermo Scientific Acros
388371000 |
100 mL | AcroSeal™ Glass Bottle |
£61.40
100mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10295983
|
Thermo Scientific Acros
388378000 |
800 mL | AcroSeal™ Glass bottle |
£306.00
800mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Trimethylamine | |
1M solution in THF | |
C3H9N | |
MFCD00008327 | |
0.8500g/mL | |
Solubility in water: soluble. Other solubilities: soluble in most organic solvents | |
CN(C)C | |
1146 | |
59.11 | |
−17°C |
Colorless to Yellow | |
75-50-3, 109-99-9 | |
N(CH3)3 | |
1M, exact strength on the certificate of analysis | |
trimethylamine, methanamine, n,n-dimethyl, dimethylmethaneamine, n-trimethylamine, trimethylamine solution, ch3 3n, trimethyl amine, trimethylamin, trimethyl-amine, fema number 3241 | |
GETQZCLCWQTVFV-UHFFFAOYSA-N | |
59.11 | |
CHEBI:18139 | |
0.85 | |
Liquid |
Safety and Handling
GHS H Statement
Highly flammable liquid and vapor. Causes serious eye damage. May cause respiratory irritation. Causes skin irritation. Suspected of causing cancer. Harmful if swallowed. May cause drowsiness or dizziness. May form explosive peroxides.
GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces. - No smoking. IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/shower. IF SWALLOWED: rinse mouth. Do NOT induce vomiting. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Immediately call a POISON CENTER or doctor/physician. IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing. Wear protective gloves/protective clothing/eye protection/face protection.
Danger
- Trimethylamine, 1M solution in THF
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 4
- Carcinogenicity Category 2
- Serious eye damage/eye irritation Category 1
- Flammable liquid Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H318-Causes serious eye damage.
- H315-Causes skin irritation.
- H351-Suspected of causing cancer.
- H335-May cause respiratory irritation.
- H336-May cause drowsiness or dizziness.
- H302-Harmful if swallowed.
- H225-Highly flammable liquid and vapour.
- EUH019-May form explosive peroxides.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only