missing translation for 'onlineSavingsMsg'
Learn More

Trimethylhydroquinone, 97%, Thermo Scientific Chemicals

Catalog Number 10357120
Click to view available options
Quantity:
100 g
500 g
Packaging:
Plastic bottle
This item is not returnable. View return policy
700-13-0
C9H12O2
152.19
MFCD00002346
AUFZRCJENRSRLY-UHFFFAOYSA-N
trimethylhydroquinone, 2,3,5-trimethylhydroquinone, 1,4-benzenediol, 2,3,5-trimethyl, pseudocumohydroquinone, psi-cumohydroquinone, 2,3,6-trimethylhydroquinone, 2,3,5-trimethyl-benzene-1,4-diol, hydroquinone, trimethyl, 2,3,5-trimethyl-1,4-benzenediol, .psi.-cumohydroquinone
12785
2,3,5-trimethylbenzene-1,4-diol
CC1=CC(=C(C(=C1O)C)C)O
This item is not returnable. View return policy
700-13-0
C9H12O2
152.19
MFCD00002346
AUFZRCJENRSRLY-UHFFFAOYSA-N
trimethylhydroquinone, 2,3,5-trimethylhydroquinone, 1,4-benzenediol, 2,3,5-trimethyl, pseudocumohydroquinone, psi-cumohydroquinone, 2,3,6-trimethylhydroquinone, 2,3,5-trimethyl-benzene-1,4-diol, hydroquinone, trimethyl, 2,3,5-trimethyl-1,4-benzenediol, .psi.-cumohydroquinone
12785
2,3,5-trimethylbenzene-1,4-diol
CC1=CC(=C(C(=C1O)C)C)O

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 169°C to 174°C
Color Beige
Boiling Point 295°C
Flash Point 191°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Plastic bottle
Quantity 100 g
Beilstein 06, IV, 5997
Solubility Information Solubility in water: 2g/L water (20°C). Other solubilities: soluble in most common organic solvents
Formula Weight 152.19
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material Trimethylhydroquinone, 97%
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Trimethylhydroquinone
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Serious eye damage/eye irritation Category 1
  • Respiratory sensitiser Category 1
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H318-Causes serious eye damage.
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
  • H315-Causes skin irritation.
  • H335-May cause respiratory irritation.
  • H332-Harmful if inhaled.
Precautionary Statement
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'