missing translation for 'onlineSavingsMsg'
Learn More

Trimethylhydroquinone, 98%, Thermo Scientific Chemicals

£31.86 - £94.07

Chemical Identifiers

CAS 700-13-0
Molecular Formula C9H12O2
Molecular Weight (g/mol) 152.193
MDL Number MFCD00002346
InChI Key AUFZRCJENRSRLY-UHFFFAOYSA-N
Synonym trimethylhydroquinone, 2,3,5-trimethylhydroquinone, 1,4-benzenediol, 2,3,5-trimethyl, pseudocumohydroquinone, psi-cumohydroquinone, 2,3,6-trimethylhydroquinone, 2,3,5-trimethyl-benzene-1,4-diol, hydroquinone, trimethyl, 2,3,5-trimethyl-1,4-benzenediol, .psi.-cumohydroquinone
PubChem CID 12785
IUPAC Name 2,3,5-trimethylbenzene-1,4-diol
SMILES CC1=CC(=C(C(=C1O)C)C)O
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11429857
View Documents
Thermo Scientific Alfa Aesar
B22819.22
100 g
£31.86
100g
Please to purchase this item. Need a web account? Register with us today!
11439857
View Documents
Thermo Scientific Alfa Aesar
B22819.36
500 g
£94.07
500g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Trimethylhydroquinone is used for synthesizing vitamin E and used as medical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Trimethylhydroquinone is used for synthesizing vitamin E and used as medical intermediate.

Solubility
Soluble in water (2 g/L @ 20°C.

Notes
Store in cool, dry conditions in a sealed container. Incompatible with oxidizing agents.
Specifications

Chemical Identifiers

700-13-0
152.193
AUFZRCJENRSRLY-UHFFFAOYSA-N
12785
CC1=CC(=C(C(=C1O)C)C)O
C9H12O2
MFCD00002346
trimethylhydroquinone, 2,3,5-trimethylhydroquinone, 1,4-benzenediol, 2,3,5-trimethyl, pseudocumohydroquinone, psi-cumohydroquinone, 2,3,6-trimethylhydroquinone, 2,3,5-trimethyl-benzene-1,4-diol, hydroquinone, trimethyl, 2,3,5-trimethyl-1,4-benzenediol, .psi.-cumohydroquinone
2,3,5-trimethylbenzene-1,4-diol

Specifications

700-13-0
140°C (284°F)
MFCD00002346
1909183
AUFZRCJENRSRLY-UHFFFAOYSA-N
2,3,5-trimethylbenzene-1,4-diol
12785
98%
169°C to 174°C
C9H12O2
UN3077
trimethylhydroquinone, 2,3,5-trimethylhydroquinone, 1,4-benzenediol, 2,3,5-trimethyl, pseudocumohydroquinone, psi-cumohydroquinone, 2,3,6-trimethylhydroquinone, 2,3,5-trimethyl-benzene-1,4-diol, hydroquinone, trimethyl, 2,3,5-trimethyl-1,4-benzenediol, .psi.-cumohydroquinone
CC1=CC(=C(C(=C1O)C)C)O
152.193
152.2
Trimethylhydroquinone
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H318-H332-H317-H335-H315
Causes serious eye damage.
Harmful if inhaled.
May cause an allergic skin reaction.
May cause respiratory irritation.
Causes skin irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

missing translation for 'dotInformation' : Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.

missing translation for 'rtecsNumber' : MX7820000

missing translation for 'tsca' : TSCA

Product Identifier
  • Trimethylhydroquinone

Signal Word
  • Danger

Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 2
  • Skin sensitiser Category 1
  • Specific target organ toxicity Category 3

Hazard Statement
  • H315-Causes skin irritation.
  • H317-May cause an allergic skin reaction.
  • H318-Causes serious eye damage.
  • H332-Harmful if inhaled.
  • H335-May cause respiratory irritation.
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.

Precautionary Statement
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

Supplemental information
  • MIXTURE LIST-Contains : Trimethylhydroquinon

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only