Learn More
Trimethyloxonium tetrafluoroborate, 95%, Thermo Scientific Chemicals
£43.70 - £66.00
Chemical Identifiers
CAS | 420-37-1 |
---|---|
Molecular Formula | C3H9BF4O |
Molecular Weight (g/mol) | 147.91 |
MDL Number | MFCD00011798 |
InChI Key | CZVZBKHWOFJNCR-UHFFFAOYSA-N |
Synonym | trimethyloxonium tetrafluoroborate, trimethyloxidanium tetrafluoroborate, tetrafluoroboranuide; trimethyloxidanium, trimethyloxonium fluoborate, acmc-209jn1, ksc236c1n, trimethyloxonium tetrafluroborate, trimethyloxonium tetrafloroborate, trimethyloxonium tetrafluorborate, trimethyloxonium tetraflouroborate |
PubChem CID | 2735153 |
IUPAC Name | trimethyloxidanium;tetrafluoroborate |
SMILES | C[O+](C)C.F[B-](F)(F)F |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10311361
|
Thermo Scientific Acros
421960025 |
2.5 g | Glass bottle |
£43.70
2.50g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10750682
|
Thermo Scientific Acros
421960100 |
10 g | Glass bottle |
£66.00
10g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Alkylation reagentChemical Identifiers
420-37-1 | |
147.91 | |
CZVZBKHWOFJNCR-UHFFFAOYSA-N | |
2735153 | |
C[O+](C)C.F[B-](F)(F)F |
C3H9BF4O | |
MFCD00011798 | |
trimethyloxonium tetrafluoroborate, trimethyloxidanium tetrafluoroborate, tetrafluoroboranuide; trimethyloxidanium, trimethyloxonium fluoborate, acmc-209jn1, ksc236c1n, trimethyloxonium tetrafluroborate, trimethyloxonium tetrafloroborate, trimethyloxonium tetrafluorborate, trimethyloxonium tetraflouroborate | |
trimethyloxidanium;tetrafluoroborate |
Specifications
420-37-1 | |
White | |
C3H9BF4O | |
MFCD00011798 | |
01,1232; 03,438; 04,314; 05,541; 13,327 | |
trimethyloxonium tetrafluoroborate, trimethyloxidanium tetrafluoroborate, tetrafluoroboranuide; trimethyloxidanium, trimethyloxonium fluoborate, acmc-209jn1, ksc236c1n, trimethyloxonium tetrafluroborate, trimethyloxonium tetrafloroborate, trimethyloxonium tetrafluorborate, trimethyloxonium tetraflouroborate | |
C[O+](C)C.F[B-](F)(F)F | |
147.91 | |
147.91 | |
Crystals |
200°C | |
95% | |
(CH3)3OBF4 | |
3261 | |
15, 9889 | |
CZVZBKHWOFJNCR-UHFFFAOYSA-N | |
trimethyloxidanium;tetrafluoroborate | |
2735153 | |
95% | |
Trimethyloxonium tetrafluoroborate |
Safety and Handling
GHS H Statement
Causes severe skin burns and eye damage.
Reacts violently with water.
GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsi
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 206-994-4
- Trimethyloxonium tetrafluoroborate
Signal Word
- Danger
Hazard Category
- Serious eye damage/eye irritation Category 1
- Skin corrosion/irritation Category 1 B
Hazard Statement
- H318-Causes serious eye damage.
- H314-Causes severe skin burns and eye damage.
- EUH014-Reacts violently with water.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- P402+P404-Store in a dry place. Store in a closed container.
RUO – Research Use Only