missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Trimethylsilanol, 95%, Thermo Scientific Chemicals
Click to view available options
Quantity:
5 g
25 g
100 g
Description
Trimethylsilanol used as a hydrophobic coat on silicate surfaces. Used as an as an antimicrobial agent.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Trimethylsilanol used as a hydrophobic coat on silicate surfaces. Used as an as an antimicrobial agent.
Solubility
Soluble in water (1.00 mg/ml).
Notes
Store in cool, dry place in tightly closed container. With good ventilation. Store away from oxidizing agent.
Trimethylsilanol used as a hydrophobic coat on silicate surfaces. Used as an as an antimicrobial agent.
Solubility
Soluble in water (1.00 mg/ml).
Notes
Store in cool, dry place in tightly closed container. With good ventilation. Store away from oxidizing agent.
Specifications
Boiling Point | 98°C to 100°C |
Quantity | 100 g |
Linear Formula | (CH3)3SiOH |
UN Number | UN1993 |
Beilstein | 1361356 |
Flash Point | 4°C (39°F) |
Solubility Information | Soluble in water (1.00mg/ml). |
Formula Weight | 90.2 |
Percent Purity | 95% |
Density | 0.814 |
Show More |
Safety and Handling
Signal Word
- Danger
- Acute toxicity Category 4
- LONG-TERM AQUATIC HAZARD Chronic 3
- Flammable liquid Category 2
- H225-Highly flammable liquid and vapour.
- H332-Harmful if inhaled.
- H412-Harmful to aquatic life with long lasting effects.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
- P403+P233-Store in a well-ventilated place. Keep container tightly closed.
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'