missing translation for 'onlineSavingsMsg'
Learn More

Trimethylsilyl 2,2-difluoro-2-(fluorosulfonyl)acetate, 94%

Product Code. p-7029401
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
This item is not returnable. View return policy

Product Code. 15447625

Brand: Thermo Scientific Alfa Aesar H26857.03

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 120801-75-4 | C5H9F3O4SSi | 250.26 g/mol

Trimethylsilyl 2,2-difluoro-2-(fluorosulfonyl)acetate is an fluoroalkylation agent, reactant for preparation of difluorocyclopropenes, preparation of difluorocarbene reagents.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Trimethylsilyl 2,2-difluoro-2-(fluorosulfonyl)acetate is an fluoroalkylation agent, reactant for preparation of difluorocyclopropenes, preparation of difluorocarbene reagents.

Solubility
Reacts with water.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 120801-75-4
Molecular Formula C5H9F3O4SSi
Molecular Weight (g/mol) 250.26
MDL Number MFCD02093343
InChI Key XHVSCKNABCCCAC-UHFFFAOYSA-N
PubChem CID 2778022
IUPAC Name trimethylsilyl 2,2-difluoro-2-fluorosulfonylacetate
SMILES C[Si](C)(C)OC(=O)C(F)(F)S(F)(=O)=O

Specifications

Density 1.27
Boiling Point 156°C
Flash Point 26°C (79°F)
Refractive Index 1.367
Quantity 1 g
UN Number UN2920
Synonym trimethylsilyl 2,2-difluoro-2-fluorosulfonyl acetate, trimethylsilyl 2-fluorosulfonyl difluoroacetate, trimethylsilyl2-fluorosulphonyl difluoroacetate, trimethylsilyl fluorosulfonyldifluoroacetate, trimethylsilyl difluoro fluorosulfonyl acetate, trimethylsilyl 2-fluorosulphonyl difluoroacetate, 2,2-difluoro-2-fluorosulfonyl acetic acid trimethylsilyl ester, pubchem22642, g00021-watson-int, acmc-1c7yr
Solubility Information Reacts with water.
Formula Weight 250.27
Percent Purity 94%
Chemical Name or Material Trimethylsilyl 2,2-difluoro-2-(fluorosulfonyl)acetate
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.