Learn More
(Trimethylsilyl)diazomethane, 2M in hexanes, Thermo Scientific Chemicals
£67.10 - £781.00
Chemical Identifiers
CAS | 18107-18-1 |
---|---|
Molecular Formula | C4H10N2Si |
Molecular Weight (g/mol) | 114.223 |
MDL Number | MFCD00053946 |
InChI Key | ONDSBJMLAHVLMI-UHFFFAOYSA-N |
Synonym | trimethylsilyl diazomethane, trimethylsilyldiazomethane, diazomethyl trimethylsilane, tms-diazomethane, diazomethyl trimethyl silane, tmschn2, silane, diazomethyl trimethyl, unii-qi98hqo8c4, diazomethyl-trimethyl-silane, qi98hqo8c4 |
PubChem CID | 167693 |
IUPAC Name | diazomethyl(trimethyl)silane |
SMILES | C[Si](C)(C)C=[N+]=[N-] |
Description
(Trimethylsilyl)diazomethane is used as a methylating agent in organic chemistry. It is utilized in Gas Chromatography- Mass-Spectrometry (GC-MS) for the analysis of various carboxylic compounds. It is involved as a reagent in the Doyle-Kirmse reaction with allyl sulfides and allyl amines. It is considered and used as a less-explosive replacement fordiazomethane. It is also used as a reactant for Unnatural alfa-amino acid derivatives containing gem-bisphosphonates through Michael addition reaction, imidazolo[2,1-b]benzothiazole derivatives and aigialomycin D analogues.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications(Trimethylsilyl)diazomethane is used as a methylating agent in organic chemistry. It is utilized in Gas Chromatography- Mass-Spectrometry (GC-MS) for the analysis of various carboxylic compounds. It is involved as a reagent in the Doyle-Kirmse reaction with allyl sulfides and allyl amines. It is considered and used as a less-explosive replacement fordiazomethane. It is also used as a reactant for Unnatural alfa-amino acid derivatives containing gem-bisphosphonates through Michael addition reaction, imidazolo[2,1-b]benzothiazole derivatives and aigialomycin D analogues.
Solubility
Immiscible with water. Miscible with organic solvents.
Notes
(Trimethylsilyl)diazomethane (TMSD) in alcoholic solvents under acidic or basic conditions can lead to the formation of diazomethane. TMSD is under review by the National Toxicological Program for potential inhalation hazards.Incompatible with oxidizing agents, chlorine, fluorine and perchlorates.
Chemical Identifiers
18107-18-1 | |
114.223 | |
ONDSBJMLAHVLMI-UHFFFAOYSA-N | |
167693 | |
C[Si](C)(C)C=[N+]=[N-] |
C4H10N2Si | |
MFCD00053946 | |
trimethylsilyl diazomethane, trimethylsilyldiazomethane, diazomethyl trimethylsilane, tms-diazomethane, diazomethyl trimethyl silane, tmschn2, silane, diazomethyl trimethyl, unii-qi98hqo8c4, diazomethyl-trimethyl-silane, qi98hqo8c4 | |
diazomethyl(trimethyl)silane |
Specifications
18107-18-1 | |
MFCD00053946 | |
1902903 | |
trimethylsilyl diazomethane, trimethylsilyldiazomethane, diazomethyl trimethylsilane, tms-diazomethane, diazomethyl trimethyl silane, tmschn2, silane, diazomethyl trimethyl, unii-qi98hqo8c4, diazomethyl-trimethyl-silane, qi98hqo8c4 | |
ONDSBJMLAHVLMI-UHFFFAOYSA-N | |
diazomethyl(trimethyl)silane | |
167693 | |
2M in hexanes | |
0.718 |
C4H10N2Si | |
UN3383 | |
−23°C (−10°F) | |
Immiscible with water. Miscible with organic solvents. | |
C[Si](C)(C)C=[N+]=[N-] | |
114.223 | |
114.22 | |
Light and moisture sensitive | |
(Trimethylsilyl)diazomethane |
Safety and Handling
GHS H Statement
H225-H331-H350-H304-H361-H373-H315-H336
Highly flammable liquid and vapor.
Toxic if inhaled.
May cause cancer.
May be fatal if swallowed and enters airways.
Suspected of damaging fertility or the unborn child.
May cause damage to organs through prolonged or repeated exposure.
Causes skin irritation.
May cause drowsiness or dizziness.
P201-P202-P210-P233-P235-P240-P241-P242-P243-P260-P264b-P271-P280i-P281-P284-P301+P310-P303+P361+P353-P304+P340-P305+P351+P338-P308+P313-P310-P331-P332+P313-P363-P370+P378q-P5
H225-H304-H315-H319-H330-H335-H336-H350-H361f-H373
missing translation for 'dotInformation' : Hazard Class: 3; Packaging Group: II
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures; Store under Nitrogen
RUO – Research Use Only