missing translation for 'onlineSavingsMsg'
Learn More

(Trimethylsilyl)methylmagnesium chloride, 0.5M in 2-MeTHF, Thermo Scientific Chemicals

Product Code. 15451426
Click to view available options
Quantity:
100 mL
This item is not returnable. View return policy
This item is not returnable. View return policy

 

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 13170-43-9
Molecular Formula C4H11ClMgSi
Linear Formula (CH3)3SiCH2MgCl
MDL Number MFCD00009916
Quantity 100 mL
UN Number UN2924
Sensitivity Air and moisture sensitive
Synonym trimethylsilylmethylmagnesium chloride, tmsch2mgcl, trimethylsilyl methylmagnesium chloride, trimethylsilyl methyl magnesium chloride, trimethylsilyl methylmagnesium chloride solution, me3sich2mgcl, naqatmjwcjchoz-uhfffaoysa-m, chlorotrimethylsilylmethyl magnesium, trimethylsiylmethylmagnesium chloride
InChI Key NAQATMJWCJCHOZ-UHFFFAOYSA-M
SMILES C[Si](C)(C)C[Mg]Cl
IUPAC Name magnesium;methanidyl(trimethyl)silane;chloride
Molecular Weight (g/mol) 146.97
PubChem CID 2734900
Formula Weight 146.97
Concentration or Composition (by Analyte or Components) 0.5M in 2-MeTHF
Chemical Name or Material (Trimethylsilyl)methylmagnesium chloride
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'