missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Trimethylsilyl trifluoromethanesulfonate, 99%, Thermo Scientific Chemicals
Click to view available options
Quantity:
10 mL
50 mL
250 mL
Packaging:
Glass bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
Color | Colorless to Brown |
Density | 1.2250g/mL |
Boiling Point | 140°C |
Flash Point | 25°C |
Infrared Spectrum | Authentic |
Assay Percent Range | 99% |
Packaging | Glass bottle |
Linear Formula | CF3SO3Si(CH3)3 |
Refractive Index | 1.3600 to 1.362 |
Quantity | 250 mL |
Show More |
Safety and Handling
Product Identifier
- Trimethylsilyl trifluoromethanesulfonate
- Danger
- Flammable liquid Category 3
- Skin corrosion/irritation Category 1 B
- Serious eye damage/eye irritation Category 1
- H226-Flammable liquid and vapour.
- H314-Causes severe skin burns and eye damage.
- EUH014-Reacts violently with water.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- P402+P404-Store in a dry place. Store in a closed container.
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'