Learn More
Trimethyltin chloride, Thermo Scientific Chemicals
£31.40 - £301.00
Chemical Identifiers
CAS | 1066-45-1 |
---|---|
Molecular Formula | C3H11ClSn |
Molecular Weight (g/mol) | 201.28 |
MDL Number | MFCD00000520 |
InChI Key | FVFGWISLDLUGHX-UHFFFAOYSA-N |
Synonym | trimethyltin chloride, stannane, chlorotrimethyl, chlorotrimethyltin, trimethylchlorotin, trimethylstannyl chloride, trimethylchlorostannane, chloro trimethyl stannane, m&t chemicals 1222-45, stannylium, trimethyl-, chloride, trimethyltinchloride |
PubChem CID | 14016 |
IUPAC Name | chloro(trimethyl)stannane |
SMILES | Cl.C[SnH](C)C |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11334859
|
Thermo Scientific Alfa Aesar
071166.04 |
2 g |
£31.40
2g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11324859
|
Thermo Scientific Alfa Aesar
071166.09 |
10 g | N/A | ||||||
11344859
|
Thermo Scientific Alfa Aesar
071166.18 |
50 g |
£301.00
50g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Trimethyltin chloride is used as a heat stabilizer in the manufacture of vinyl window blinds. It is also used as a stabilizer for plastics and as additive to paint, as antifouling agent. It also serves as a byproduct of methyltin manufacture. Further, it is used as a precursor to vinyltrimethylstannane and in polyvinyl chloride stabilization.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
1066-45-1 | |
201.28 | |
FVFGWISLDLUGHX-UHFFFAOYSA-N | |
14016 | |
Cl.C[SnH](C)C |
C3H11ClSn | |
MFCD00000520 | |
trimethyltin chloride, stannane, chlorotrimethyl, chlorotrimethyltin, trimethylchlorotin, trimethylstannyl chloride, trimethylchlorostannane, chloro trimethyl stannane, m&t chemicals 1222-45, stannylium, trimethyl-, chloride, trimethyltinchloride | |
chloro(trimethyl)stannane |
Specifications
1066-45-1 | |
White | |
97°C (207°F) | |
C3H11ClSn | |
UN3146 | |
trimethyltin chloride, stannane, chlorotrimethyl, chlorotrimethyltin, trimethylchlorotin, trimethylstannyl chloride, trimethylchlorostannane, chloro trimethyl stannane, m&t chemicals 1222-45, stannylium, trimethyl-, chloride, trimethyltinchloride | |
FVFGWISLDLUGHX-UHFFFAOYSA-N | |
chloro(trimethyl)stannane | |
14016 | |
Crystalline |
37°C to 39°C | |
154°C | |
Unpleasant | |
MFCD00000520 | |
Moisture sensitive | |
Soluble in water,chloroform and organic solvents. | |
Cl.C[SnH](C)C | |
201.28 | |
199.25 | |
Trimethyltin chloride |
Safety and Handling
GHS H Statement
H300-H310-H330
Fatal if swallowed.
Fatal in contact with skin.
Fatal if inhaled.
P260-P262-P264b-P270-P271-P280-P284-P301+P310-P302+P350-P304+P340-P310-P312-P314-P330-P361-P363-P501c
H300+H310+H330-H335-H336-H373
missing translation for 'dotInformation' : Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: ORGANOTIN COMPOUNDS, SOLID, N.O.S.
missing translation for 'einecsNumber' : 213-917-8
missing translation for 'rtecsNumber' : WH6850000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures; Store under Nitrogen
RUO – Research Use Only