Learn More
Trimethyltin hydroxide, 98%, Thermo Scientific Chemicals
Description
Trimethyltin hydroxide is used in the synthesis of modified thymines, which is useful as inhibitors of RNase A. It is also used in the preparation of ketoheterocycle inhibitors of fatty acid amide inhibitors. It reacts with 2,2-disubstituted benzothiazolines to form trialkyltin derivatives of sulphur containing Schiff base complexes.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
Melting Point | 114°C to 118°C |
Color | White |
Odor | Pungent |
Linear Formula | (CH3)3SnOH |
Quantity | 1 g |
UN Number | UN3146 |
Sensitivity | Moisture sensitive |
Solubility Information | Soluble in water. |
Formula Weight | 180.8 |
Percent Purity | 98% |
Show More |
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
U15J097 | Certificate of Analysis | 24/10/2023 | 071167.06,071167.03,071167.14 |
T13J053 | Certificate of Analysis | 19/07/2023 | 071167.06,071167,071167.03,071167.14 |
Z30H026 | Certificate of Analysis | 12/04/2022 | 071167.06,071167.03,071167.14 |
X30F036 | Certificate of Analysis | 28/12/2021 | 071167.06,071167.03,071167.14 |
S04A048 | Certificate of Analysis | 08/12/2021 | 071167.06,071167.03,071167.14 |
Safety and Handling


- Trimethyltin hydroxide
- Danger
- Acute toxicity Category 1
- Acute toxicity Category 2
- LONG-TERM AQUATIC HAZARD Chronic 1
- ACUTE AQUATIC Acute 1
- H300-Fatal if swallowed.
- H310-Fatal in contact with skin.
- H330-Fatal if inhaled.
- H400-Very toxic to aquatic life.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P273-Avoid release to the environment.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- MIXTURE LIST-Contains : Trimethyltin hydroxide
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.