Learn More
Olympus™ Trinocular Microscope, Basic Stand, No Illumination, SZ61
Deliver images with superb depth, clarity, and detail. Olympus™ Trinocular Microscope, Basic Stand, No Illumination, SZ61 uses dependable, high-performance optics to produce consistent and accurate results and a V-shape optical path to ensure a slim body ideal for equipment integration or for stand alone use.
Brand: Olympus™
Code : NEW
Additional Details : Weight : 6.50000kg
Description
- Trinocular head, 45º inclination.
- 0.5X C-Mount camera adapter for 1/2″ sensor camera.
- Interpupillary adjustment 52-76mm.
- Greenough zoom optical system.
- Widefield eyepieces WF10X FN22mm.
- Zoom ratio 6.7:1.
- 110mm working distance.
- Stand with focusing & ESD capability.
- Glass stage plate.
- Dust Cover + PSU.
Specifications
WF 10X FN22 mm | |
Microscope | |
Trinocular | |
0.67X to 4.5X |