missing translation for 'onlineSavingsMsg'
Learn More

Trioctyl phosphate

Product Code. 11387138
Click to view available options
Quantity:
50 g
250 g
1 kg
Unit Size:
1kg
250g
50g
This item is not returnable. View return policy

Product Code. 11387138

Brand: Thermo Scientific Alfa Aesar 015808.30

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 78-42-2 | C24H51O4P | 434.64 g/mol

Trioctyl phosphate finds application as a fire-retardant, plasticizer for polyvinyl chloride and cellulose nitrate. It is used as a stimulant for the chemical warfare nerve agent VX. Further, it used as an alkylation agent for nitrogen heterocyclic compounds and as a catalyst to produce phenolic and urea resins. In addition to this, it is used as an antifoaming agent and a co-solvent in hydrogen peroxide production.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 78-42-2
Molecular Formula C24H51O4P
Molecular Weight (g/mol) 434.64
MDL Number MFCD00009491
InChI Key GTVWRXDRKAHEAD-UHFFFAOYNA-N
Synonym tris 2-ethylhexyl phosphate, disflamoll tof, flexol tof, phosphoric acid, tris 2-ethylhexyl ester, kronitex tof, flexol plasticizer tof, 2-ethyl-1-hexanol phosphate, tris ethylhexyl phosphate, tri 2-ethylhexyl phosphate
PubChem CID 6537
IUPAC Name tris(2-ethylhexyl) phosphate
SMILES CCCCC(CC)COP(=O)(OCC(CC)CCCC)OCC(CC)CCCC

Specifications

Density 0.93 g/mL
Boiling Point 216°C (4 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.441
Quantity 250 g
Solubility Information Miscible with mineral oil and gasoline. Immiscible with water.
Formula Weight 434.64
Physical Form Liquid
Chemical Name or Material Trioctyl phosphate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.