missing translation for 'onlineSavingsMsg'
Learn More

Corning™ Stripettor™ Ultra Pipet Controller Accessory, Colored Nose Pieces (Set)

For use with Stripettor™ Ultra Pipet Controller

Brand:  Corning™ 4997

Additional Details : Weight : 0.30100kg

Product Code. 17780708

  • £11.33 / Each
Estimated Shipment: 22-11-2024
to see stock.
Add to basket

Description

Description

Color coded for easy identification
Specifications

Specifications

Set of Colored Nose pieces
Ultra Pipet Controller
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers