missing translation for 'onlineSavingsMsg'
Learn More

Triphenylarsine, 97%, Thermo Scientific Chemicals

Catalog Number 11372236
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
603-32-7
C18H15As
306.24
MFCD00002994
BPLUKJNHPBNVQL-UHFFFAOYSA-N
triphenylarsine, arsine, triphenyl, triphenyl arsenic, triphenylarsene, arsinetriphenyl, unii-mn8ez3fl74, mn8ez3fl74, triphenylarsenic, triphenyl arsine, triphenyl-arsine
11773
triphenylarsane
C1=CC=C(C=C1)[As](C2=CC=CC=C2)C3=CC=CC=C3
This item is not returnable. View return policy
603-32-7
C18H15As
306.24
MFCD00002994
BPLUKJNHPBNVQL-UHFFFAOYSA-N
triphenylarsine, arsine, triphenyl, triphenyl arsenic, triphenylarsene, arsinetriphenyl, unii-mn8ez3fl74, mn8ez3fl74, triphenylarsenic, triphenyl arsine, triphenyl-arsine
11773
triphenylarsane
C1=CC=C(C=C1)[As](C2=CC=CC=C2)C3=CC=CC=C3

Triphenylarsine is used in certain metal-catalyzed coupling reactions. It is a precursor to tetraphenylarsonium chloride a popular precipitating agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Triphenylarsine is used in certain metal-catalyzed coupling reactions. It is a precursor to tetraphenylarsonium chloride a popular precipitating agent.

Solubility
Soluble in methanol. Insoluble in water.

Notes
Handle and store under inert gas. Incompatible materials are Strong oxidizing agents.
TRUSTED_SUSTAINABILITY
Melting Point 58°C to 62°C
Boiling Point 360°C
Flash Point 265°C (509°F)
Linear Formula (C6H5)3As
Quantity 5 g
UN Number UN3465
Beilstein 1842037
Solubility Information Soluble in methanol. Insoluble in water.
Formula Weight 306.24
Percent Purity 97%
Chemical Name or Material Triphenylarsine
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'