missing translation for 'onlineSavingsMsg'
Learn More

Triphenylmethane, 98%, Thermo Scientific Chemicals

Product Code. 11449223
Click to view available options
Quantity:
25 g
100 g
500 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 519-73-3
Molecular Formula C19H16
Molecular Weight (g/mol) 244.34
MDL Number MFCD00004763
InChI Key AAAQKTZKLRYKHR-UHFFFAOYSA-N
Synonym triphenylmethane, tritane, benzene, 1,1',1-methylidynetris, methane, triphenyl, diphenylmethyl benzene, 1,1',1-methanetriyltribenzene, unii-8o4utw9e17, ccris 5194, triphenyl methane, triphenylmethane?
PubChem CID 10614
ChEBI CHEBI:76212
IUPAC Name benzhydrylbenzene
SMILES C1=CC=C(C=C1)C(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 92°C to 94°C
Density 1.01
Boiling Point 358°C to 359°C
Quantity 500 g
Beilstein 1909753
Merck Index 14,9741
Formula Weight 244.34
Percent Purity 98%
Chemical Name or Material Triphenylmethane

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'