Learn More
Chemical Identifiers
CAS | 76-84-6 |
---|---|
Molecular Formula | C19H16O |
Molecular Weight (g/mol) | 260.34 |
MDL Number | MFCD00004445,MFCD10565638 |
InChI Key | LZTRCELOJRDYMQ-UHFFFAOYSA-N |
Synonym | triphenylcarbinol, trityl alcohol, tritanol, triphenylmethyl alcohol, triphenyl methanol, triphenyl carbinol, methanol, triphenyl, unii-u97q0ou9kb, triphenyl-methanol, benzenemethanol, .alpha.,.alpha.-diphenyl |
PubChem CID | 6457 |
IUPAC Name | triphenylmethanol |
SMILES | OC(C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11444173
|
Thermo Scientific Alfa Aesar
A10366.18 |
50 g |
£25.93
50g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11454173
|
Thermo Scientific Alfa Aesar
A10366.30 |
250 g |
£84.02
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11434173
|
Thermo Scientific Alfa Aesar
A10366.0B |
1000 g |
£274.37
1000g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Triphenylmethanol is used as a reagent in the research laboratory. It acts as an intermediate in the production of the commercially useful triarylmethane dyes. It is used in the preparation of triphenylmethane. It is also used as an antiproliferative agent. Further, it is used in the preparation of two-electron reduction product of pyrylogen. In addition to this, it reacts with triphenylphosphine oxide to form a 1:1 molecular complex. It serves as a specific clathrate host for methanol and dimethyl sulphoxide and forms clathrate inclusion complexes.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsTriphenylmethanol is used as a reagent in the research laboratory. It acts as an intermediate in the production of the commercially useful triarylmethane dyes. It is used in the preparation of triphenylmethane. It is also used as an antiproliferative agent. Further, it is used in the preparation of two-electron reduction product of pyrylogen. In addition to this, it reacts with triphenylphosphine oxide to form a 1:1 molecular complex. It serves as a specific clathrate host for methanol and dimethyl sulphoxide and forms clathrate inclusion complexes.
Solubility
Soluble in dioxane, ethanol, water, ether and benzene.
Notes
Incompatible with acids, acid chlorides, acid anhydrides and oxidizing agents.
Chemical Identifiers
76-84-6 | |
260.34 | |
LZTRCELOJRDYMQ-UHFFFAOYSA-N | |
6457 | |
OC(C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1 |
C19H16O | |
MFCD00004445,MFCD10565638 | |
triphenylcarbinol, trityl alcohol, tritanol, triphenylmethyl alcohol, triphenyl methanol, triphenyl carbinol, methanol, triphenyl, unii-u97q0ou9kb, triphenyl-methanol, benzenemethanol, .alpha.,.alpha.-diphenyl | |
triphenylmethanol |
Specifications
76-84-6 | |
360°C to 380°C | |
C19H16O | |
1460837 | |
triphenylcarbinol, trityl alcohol, tritanol, triphenylmethyl alcohol, triphenyl methanol, triphenyl carbinol, methanol, triphenyl, unii-u97q0ou9kb, triphenyl-methanol, benzenemethanol, .alpha.,.alpha.-diphenyl | |
LZTRCELOJRDYMQ-UHFFFAOYSA-N | |
triphenylmethanol | |
6457 | |
98% |
161°C to 164°C | |
98% | |
MFCD00004445,MFCD10565638 | |
14,9739 | |
Soluble in dioxane,ethanol,water,ether and benzene. | |
OC(C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1 | |
260.34 | |
260.34 | |
Triphenylmethanol |
Safety and Handling
GHS H Statement
H315-H319-H335
GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P362-P312-P321-P405-P403+P233-P501a
Warning
missing translation for 'einecsNumber' : 200-988-5
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only