missing translation for 'onlineSavingsMsg'
Learn More

Triphenylmethyl mercaptan, 97%, Thermo Scientific Chemicals

Product Code. 10235000
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3695-77-0
Molecular Formula C19H16S
Molecular Weight (g/mol) 276.39
MDL Number MFCD00004854
InChI Key JQZIKLPHXXBMCA-UHFFFAOYSA-N
Synonym triphenylmethyl mercaptan, trityl mercaptan, tritylthiol, triphenylmethylmercaptan, methanethiol, triphenyl, tritylmercaptan, triphenyl-methanethiol, alpha,alpha-diphenylbenzenemethanethiol, benzenemethanethiol, alpha,alpha-diphenyl, benzenemethanethiol, .alpha.,.alpha.-diphenyl
PubChem CID 77281
IUPAC Name triphenylmethanethiol
SMILES C1=CC=C(C=C1)C(C2=CC=CC=C2)(C3=CC=CC=C3)S

Specifications

Melting Point 103.0°C to 107.0°C
Color Cream to Yellow
Elemental Analysis 97.5% min.
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Packaging Glass bottle
Linear Formula (C6H5)3CSH
Quantity 100 g
Beilstein 06, I, 352
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in toluene
Formula Weight 276.39
Percent Purity 97%
Physical Form Powder
Chemical Name or Material Triphenylmethyl mercaptan
Show More Show Less
Product Identifier
  • Triphenylmethyl mercaptan

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'