Learn More
Chemical Identifiers
CAS | 603-35-0 |
---|---|
Molecular Formula | C18H15P |
Molecular Weight (g/mol) | 262.29 |
MDL Number | MFCD00003043 MFCD20489348 |
InChI Key | RIOQSEWOXXDEQQ-UHFFFAOYSA-N |
Synonym | triphenylphosphine, triphenyl phosphine, phosphine, triphenyl, triphenylphosphorus, triphenyl-phosphane, triphenylphosphide, phosphorustriphenyl, trifenylfosfin, trifenylfosfin czech, triphenylphosphine resin |
PubChem CID | 11776 |
IUPAC Name | triphenylphosphane |
SMILES | C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1 |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10143620
|
Thermo Scientific Acros
140420250 |
25 g | Plastic bottle |
£24.02
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10337120
|
Thermo Scientific Acros
140422500 |
250 g | Plastic bottle |
£49.04
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10734851
|
Thermo Scientific Acros
|
1 kg | Plastic bottle |
£151.76
1kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
13478358
|
Thermo Scientific Acros
140420050 |
5 kg | Plastic drum |
£504.51
5kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
603-35-0 | |
262.29 | |
RIOQSEWOXXDEQQ-UHFFFAOYSA-N | |
11776 | |
C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1 |
C18H15P | |
MFCD00003043 MFCD20489348 | |
triphenylphosphine, triphenyl phosphine, phosphine, triphenyl, triphenylphosphorus, triphenyl-phosphane, triphenylphosphide, phosphorustriphenyl, trifenylfosfin, trifenylfosfin czech, triphenylphosphine resin | |
triphenylphosphane |
Specifications
603-35-0 | |
White | |
181°C | |
98.5% min. (GC) | |
(C6H5)3P | |
16,759 | |
15,9916 | |
Solubility in water: insoluble. Other solubilities: soluble in glacial acetic acid,, soluble in benzene, chloroform, acetone and ccl4,, freely soluble in diethyl ether,, slightly soluble in alcohol | |
C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1 | |
262.29 | |
262.28 | |
Crystalline Powder, Crystals or Flakes |
78.5°C to 81.5°C | |
377.0°C | |
Authentic | |
C18H15P | |
MFCD00003043 MFCD20489348 | |
01,1238; 02,443; 03,317; 04,548; 05,725; 06,643; 07,403; 11,588; 12,550; 13,308; 14,60; 15,248; 16,264; 17,389 | |
triphenylphosphine, triphenyl phosphine, phosphine, triphenyl, triphenylphosphorus, triphenyl-phosphane, triphenylphosphide, phosphorustriphenyl, trifenylfosfin, trifenylfosfin czech, triphenylphosphine resin | |
RIOQSEWOXXDEQQ-UHFFFAOYSA-N | |
triphenylphosphane | |
11776 | |
99% | |
Triphenylphosphine |
Safety and Handling
GHS H Statement
May cause an allergic skin reaction.
Harmful if swallowed.
May cause damage to organs through prolonged or repeated exposure.
GHS P Statement
Wear eye protection/face protection.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
GHS Signal Word: Warning
missing translation for 'einecsNumber' : 210-036-0
missing translation for 'rtecsNumber' : SZ3500000
missing translation for 'tsca' : TSCA
- Triphenylphosphine
Signal Word
- Warning
Hazard Category
- Acute toxicity Category 4
- Respiratory sensitiser Category 1
- Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
- H317-May cause an allergic skin reaction.
- H373-May cause damage to organs.
- H302-Harmful if swallowed.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
RUO – Research Use Only