Learn More
Triphenylphosphine, flake, 99%, Thermo Scientific Chemicals
£37.90 - £414.00
Chemical Identifiers
CAS | 603-35-0 |
---|---|
Molecular Formula | C18H15P |
Molecular Weight (g/mol) | 262.29 |
MDL Number | MFCD00003043 MFCD20489348 |
InChI Key | RIOQSEWOXXDEQQ-UHFFFAOYSA-N |
Synonym | triphenylphosphine, triphenyl phosphine, phosphine, triphenyl, triphenylphosphorus, triphenyl-phosphane, triphenylphosphide, phosphorustriphenyl, trifenylfosfin, trifenylfosfin czech, triphenylphosphine resin |
PubChem CID | 11776 |
IUPAC Name | triphenylphosphane |
SMILES | C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11438036
|
Thermo Scientific Alfa Aesar
A14089.30 |
250 g |
£37.90
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11428036
|
Thermo Scientific Alfa Aesar
A14089.0B |
1000 g |
£106.00
1000g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11448036
|
Thermo Scientific Alfa Aesar
A14089.0I |
5000 g |
£414.00
5000g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
In the synthesis of organic compounds, phosphonium salts and other phosphorus compounds, and as a polymerization initiatorTriphenylphosphine is used in the synthesis of organic compounds due to its nucleophilicity and its reducing character. It is involved in the synthesis of biaryl compounds, phosphonium salts and other phosphorus compounds. As a reducing agent, it is used to prepare aromatic amines from the corresponding aromatic N-oxides. The anionic phosphine is usually isolated as the trisodium salt, which reacts with rhodium to form a complex that finds use in industrial hydroformylation reactions. It is also used to prepare Wilkinson's catalyst, RhCl(PPh3)3 useful to catalyze the hydrogenation of alkenes and tetrakis(triphenylphosphine)palladium(0) that is widely used to catalyse C-C coupling reactions in organic synthesis.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsIn the synthesis of organic compounds, phosphonium salts and other phosphorus compounds, and as a polymerization initiatorTriphenylphosphine is used in the synthesis of organic compounds due to its nucleophilicity and its reducing character. It is involved in the synthesis of biaryl compounds, phosphonium salts and other phosphorus compounds. As a reducing agent, it is used to prepare aromatic amines from the corresponding aromatic N-oxides. The anionic phosphine is usually isolated as the trisodium salt, which reacts with rhodium to form a complex that finds use in industrial hydroformylation reactions. It is also used to prepare Wilkinson′s catalyst, RhCl(PPh3)3 useful to catalyze the hydrogenation of alkenes and tetrakis(triphenylphosphine)palladium(0) that is widely used to catalyse C-C coupling reactions in organic synthesis.
Notes
Incompatible with oxidizing agents and acids.
Chemical Identifiers
603-35-0 | |
262.29 | |
RIOQSEWOXXDEQQ-UHFFFAOYSA-N | |
11776 | |
C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1 |
C18H15P | |
MFCD00003043 MFCD20489348 | |
triphenylphosphine, triphenyl phosphine, phosphine, triphenyl, triphenylphosphorus, triphenyl-phosphane, triphenylphosphide, phosphorustriphenyl, trifenylfosfin, trifenylfosfin czech, triphenylphosphine resin | |
triphenylphosphane |
Specifications
603-35-0 | |
1.200 | |
181°C (358°F) | |
C18H15P | |
610776 | |
triphenylphosphine, triphenyl phosphine, phosphine, triphenyl, triphenylphosphorus, triphenyl-phosphane, triphenylphosphide, phosphorustriphenyl, trifenylfosfin, trifenylfosfin czech, triphenylphosphine resin | |
RIOQSEWOXXDEQQ-UHFFFAOYSA-N | |
triphenylphosphane | |
11776 | |
99% | |
Triphenylphosphine |
79°C to 81°C | |
360°C | |
99% | |
MFCD00003043 MFCD20489348 | |
14,9743 | |
Insoluble in water; Soluble in benzene,acetone,carbon tetrachloride,ether,and glacial acetic acidSoluble in ether,benzene,carbon tetrachloride,glacial acetic acid,acetone,chloroform and alcohol; Insoluble in water. | |
C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1 | |
262.29 | |
262.29 | |
Flakes |
Safety and Handling
GHS H Statement
H373-H302-H317
P260-P264b-P270-P272-P280g-P301+P312-P302+P352-P314-P330-P333+P313-P363-P501c
H302-H317-H373
missing translation for 'einecsNumber' : 210-036-0
missing translation for 'rtecsNumber' : SZ3500000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only