Learn More
Triphenylphosphine oxide, 99%, Thermo Scientific Chemicals
£20.70 - £165.00
Chemical Identifiers
CAS | 791-28-6 |
---|---|
Molecular Formula | C18H15OP |
Molecular Weight (g/mol) | 278.29 |
MDL Number | MFCD00002080 MFCD03458802 |
InChI Key | FIQMHBFVRAXMOP-UHFFFAOYSA-N |
Synonym | triphenylphosphine oxide, phosphine oxide, triphenyl, triphenyl phosphorus oxide, triphenyl phosphine oxide, triphenylphosphane oxide, tppo, triphenylphosphineoxide, ph3po, diphenylphosphoroso benzene, triphenylphosphanoxid |
PubChem CID | 13097 |
ChEBI | CHEBI:36601 |
IUPAC Name | diphenylphosphorylbenzene |
SMILES | O=P(C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1 |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10447910
|
Thermo Scientific Acros
140430250 |
25 g | Glass bottle |
£20.70
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10457910
|
Thermo Scientific Acros
140431000 |
100 g | Glass bottle |
£51.80
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11935801
|
Thermo Scientific Acros
140435000 |
500 g | Glass bottle |
£165.00
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
791-28-6 | |
278.29 | |
FIQMHBFVRAXMOP-UHFFFAOYSA-N | |
13097 | |
diphenylphosphorylbenzene |
C18H15OP | |
MFCD00002080 MFCD03458802 | |
triphenylphosphine oxide, phosphine oxide, triphenyl, triphenyl phosphorus oxide, triphenyl phosphine oxide, triphenylphosphane oxide, tppo, triphenylphosphineoxide, ph3po, diphenylphosphoroso benzene, triphenylphosphanoxid | |
CHEBI:36601 | |
O=P(C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1 |
Specifications
791-28-6 | |
Brown-Pink to White | |
180°C | |
98.5% min. (HPLC) | |
(C6H5)3P(O) | |
16,IV,1011 | |
triphenylphosphine oxide, phosphine oxide, triphenyl, triphenyl phosphorus oxide, triphenyl phosphine oxide, triphenylphosphane oxide, tppo, triphenylphosphineoxide, ph3po, diphenylphosphoroso benzene, triphenylphosphanoxid | |
FIQMHBFVRAXMOP-UHFFFAOYSA-N | |
diphenylphosphorylbenzene | |
13097 | |
278.29 | |
Crystalline Powder or Flakes |
154.0°C to 158.0°C | |
>360.0°C | |
Authentic | |
C18H15OP | |
MFCD00002080 MFCD03458802 | |
14,337; 16,368 | |
Solubility in water: slightly soluble. | |
O=P(C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1 | |
278.29 | |
CHEBI:36601 | |
99% | |
Triphenylphosphine oxide |
Safety and Handling
GHS H Statement
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Harmful to aquatic life with long lasting effects.
GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
If eye irritation persists: Get medical advice/attent
GHS Signal Word: Warning
missing translation for 'einecsNumber' : 212-338-8
- Triphenylphosphine oxide
Signal Word
- Warning
Hazard Category
- Acute toxicity Category 4
- LONG-TERM AQUATIC HAZARD Chronic 3
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H412-Harmful to aquatic life with long lasting effects.
- H302-Harmful if swallowed.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
- P337+P313-If eye irritation persists: Get medical advice/attention.
RUO – Research Use Only