missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Triphenylphosphine resin, 1% crosslinked with DVB, 1.0-1.5mmol/g, 100-200 mesh, Thermo Scientific Chemicals
£87.00 - £277.00
Chemical Identifiers
CAS | 39319-11-4 |
---|---|
Molecular Formula | C18H15P |
Molecular Weight (g/mol) | 262.29 |
MDL Number | MFCD00003043 MFCD20489348 |
InChI Key | RIOQSEWOXXDEQQ-UHFFFAOYSA-N |
PubChem CID | 11776 |
IUPAC Name | triphenylphosphane |
SMILES | C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1 |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10580532
|
Thermo Scientific Acros
366870050 |
5 g | Glass bottle |
£87.00
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10025403
|
Thermo Scientific Acros
366870250 |
25 g | Glass bottle |
£277.00
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
39319-11-4 | |
262.29 | |
RIOQSEWOXXDEQQ-UHFFFAOYSA-N | |
triphenylphosphane |
C18H15P | |
MFCD00003043 MFCD20489348 | |
11776 | |
C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1 |
Specifications
39319-11-4 | |
C18H15P | |
RIOQSEWOXXDEQQ-UHFFFAOYSA-N | |
triphenylphosphane | |
11776 |
100 To 200 Mesh | |
MFCD00003043 MFCD20489348 | |
C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1 | |
262.29 | |
Triphenylphosphine resin, 1% crosslinked with DVB, 1.0-1.5mmol/g |
RUO – Research Use Only