Learn More
Triphenylphosphine resin, 1% DVB, 100-200 mesh, 1-1.5 mmol/g, Thermo Scientific™
£50.70
Chemical Identifiers
CAS | 39319-11-4 |
---|---|
Molecular Formula | C18H15P |
Molecular Weight (g/mol) | 262.29 |
MDL Number | MFCD00003043 MFCD20489348 |
InChI Key | RIOQSEWOXXDEQQ-UHFFFAOYSA-N |
PubChem CID | 11776 |
IUPAC Name | triphenylphosphane |
SMILES | C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1 |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10098771
|
Thermo Scientific Acros
358330050 |
5 g | Glass bottle | N/A | |||||
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
39319-11-4 | |
262.29 | |
RIOQSEWOXXDEQQ-UHFFFAOYSA-N | |
triphenylphosphane |
C18H15P | |
MFCD00003043 MFCD20489348 | |
11776 | |
C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1 |
Specifications
39319-11-4 | |
MFCD00003043 MFCD20489348 | |
C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1 | |
262.29 | |
Triphenylphosphine resin, Crosslinked with 1% DVB, 1.2-1.5 mmol/g, 100-200 mesh |
C18H15P | |
RIOQSEWOXXDEQQ-UHFFFAOYSA-N | |
triphenylphosphane | |
11776 |
Safety and Handling
GHS H Statement:
Harmful if swallowed.
Harmful if inhaled.
Harmful in contact with skin.
GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.