missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Triphenyltin chloride, 95%, Thermo Scientific Chemicals
Click to view available options
Quantity:
25 g
100 g
500 g
Description
Triphenyltin chloride is an important organic intermediate. It is used in agrochemical, pharmaceutical and dyestuff field.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Triphenyltin chloride is an important organic intermediate. It is used in agrochemical, pharmaceutical and dyestuff field.
Solubility
Soluble in water (0.005g/L at 20°C).
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. It is hygroscopic in nature and sensitive to moisture.
Triphenyltin chloride is an important organic intermediate. It is used in agrochemical, pharmaceutical and dyestuff field.
Solubility
Soluble in water (0.005g/L at 20°C).
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. It is hygroscopic in nature and sensitive to moisture.
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Melting Point | 103°C to 106°C |
Boiling Point | 240°C (13.5 mmHg) |
Flash Point | >110°C (230°F) |
Odor | Unpleasant |
Quantity | 25 g |
UN Number | UN3146 |
Beilstein | 524762 |
Sensitivity | Moisture sensitive |
Solubility Information | Soluble in water (0.005g/L at 20°C). |
Formula Weight | 385.48 |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'