missing translation for 'onlineSavingsMsg'
Learn More
Triphenyltin fluoride, Thermo Scientific Chemicals Product Code.: 11324527

Triphenyltin fluoride, Thermo Scientific Chemicals

Product Code. 11324527
1 g
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy

Product Code. 11324527

Brand: Thermo Scientific Alfa Aesar 039600.03

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 379-52-2
Molecular Formula C18H17FSn
Molecular Weight (g/mol) 371.04
MDL Number MFCD00015356
InChI Key SZNZEEYJVCQHCQ-UHFFFAOYSA-N
Synonym triphenyltin fluoride, fentin fluoride, stannane, fluorotriphenyl, biomet 204, tin, fluorotriphenyl, caswell no. 896h, unii-1yp2l952md, fluoro triphenyl stannane, epa pesticide chemical code 083602, triphenylzinnfluoridShow MoreShow Less
PubChem CID 9786
IUPAC Name fluoro(triphenyl)stannane
SMILES F.C1=CC=C(C=C1)[SnH](C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point >281°C
Quantity 1 g
UN Number UN3146
Formula Weight 368.99
Physical Form Powder
Chemical Name or Material Triphenyltin fluoride

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

4 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberN09I049 Certificate TypeCertificate of Analysis Date12/04/2022 Catalog Number039600.06,039600.03
Lot NumberT13D049 Certificate TypeCertificate of Analysis Date12/04/2022 Catalog Number039600.06,039600.03
Lot NumberI02T014 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog Number039600.06,039600.03
Lot NumberA18X011 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog Number039600.06,039600.03

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Triphenyltin fluoride, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.