missing translation for 'onlineSavingsMsg'
Learn More

Triphenyltin hydride, 97%, Thermo Scientific™

£44.40 - £179.00

Chemical Identifiers

CAS 892-20-6
Molecular Formula C18H16Sn
MDL Number MFCD00003004
Synonym Triphenylstannane
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11302685
View Documents
Thermo Scientific Alfa Aesar
L00163.06
5 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
11312685
View Documents
Thermo Scientific Alfa Aesar
L00163.14
25 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
Triphenyltin hydride is used as a source of “H·” to generate radicals or cleave carbon-oxygen bonds. And it is also used in anti-fouling paints.

Solubility
Insoluble in water.

Notes
Air & Moisture Sensitive. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Specifications

Chemical Identifiers

892-20-6
C18H16Sn
MFCD00003004
Triphenylstannane

Specifications

892-20-6
1.374
>110°C (230°F)
(C6H5)3SnH
MFCD00003004
3544353
Triphenylstannane
351.02
Triphenyltin hydride
26°C to 28°C
162°C to 168°C (0.5 mmHg)
C18H16Sn
1.637
UN3146
Air and moisture sensitive
Insoluble in water.
97%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H301-H311-H331
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.

P261-P264b-P270-P271-P280-P301+P310-P302+P352-P304+P340-P311-P312-P330-P361-P363-P501c

H301+H311+H331

missing translation for 'dotInformation' : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: ORGANOTIN COMPOUNDS, SOLID, N.O.S.

missing translation for 'einecsNumber' : 212-967-8

missing translation for 'rtecsNumber' : WH8882000

missing translation for 'tsca' : No

Recommended Storage : Store at -20°C

SDS
Documents

Documents

Product Certifications