missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BTX™ Triple Electroporation Tweezertrode
Triple Electroporation Tweezertrode is a reusable non-invasive, tweezer-style electrode for drug or gene delivery in animal tissues.
£4445.00 - £4635.00
Specifications
Autoclavable | Not Autoclavable |
---|---|
Type | Tweezers |
Disposable | Reusable |
For Use With (Equipment) | ECM 830, ECM 2001, Gemini X2 |
Sterility | Non-sterile |
Product Code | Brand | Diameter (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Diameter (Metric) | Price | Quantity & Availability | |||||
15741078
|
BTX™
450493 |
3 mm |
£4445.00
Each |
Estimated Shipment: 01-08-2024
Log in to see stock. |
|||||
15625028
|
BTX™
450494 |
5 mm |
£4635.00
Each |
Estimated Shipment: 03-09-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Triple Tweezertrodes have:
- Three electrode contacts with adjustable position and polarities
- Improves electrical field distribution for in utero electroporation applications
Compatability:
- ECM830
- ECM2001+
- Gemini X2
- Connect to BTX generators using the Tweezertrode Connection cables (45-0204)
Applications:
- In vivo drug or gene delivery
- Ex vivo drug or gene delivery
- In utero drug or gene delivery
Tweezertrodes may be cleaned with a mild detergent and sterilized with 70% ethanol or ethylene oxide.
Specifications
Not Autoclavable | |
Reusable | |
Non-sterile |
Tweezers | |
ECM 830, ECM 2001, Gemini X2 |