Learn More
Thermo Scientific™ Triple Wrap Sterile Pack w/VHP Indicator Sabouraud Dextrose Agar w/Lecithin, Polysorbate 80, Sodium Thiosulphate, L-Histidine
Carry out environmental monitoring in the most regulated environments with Thermo Scientific™ Triple Wrapped Irradiated plates for air sampling applications.
Brand: Thermo Scientific™ PO5513D
262.23 GBP valid until 2024-12-31
Use promo code "23263" to get your promotional price.
Additional Details : Weight : 0.33000kg
Description
Experience next-level cleanroom confidence with Thermo Scientific™ Triple Wrap Sterile Pack Sabouraud Dextrose Agar plates, complete with VHP Indicator, Lecithin, Polysorbate 80, Sodium Thiosulphate and L-Histidine.
Achieve confidence in your results and ensure optimum media fertility using plates with a Vapour-phase Hydrogen Peroxide (VHP) exposure indicator, designed to confirm the integrity of the wrapping material and seal during VHP exposure.Three protective layers reduce risk of contamination:
- Primary film with moisture control patch
- Secondary film VHP barrier with immobilized desiccant sachet
- Tertiary film dust cover
Using plates with an improved shelf life and room temperature storage offers a cost-effective solution, minimizing new batch validation and refrigeration costs
Not all products are available for sale in all territories. Please inquire.
Specifications
Ivory, transparent | |
Triple Wrap Sterile Pack w/VHP Indicator Sabouraud Dextrose Agar w/Lecithin, Polysorbate 80, Sodium Thiosulphate, L-Histidine | |
Germany | |
13.0g ± 0.5g | |
10/Pk. |
2 to 25°C | |
55mm Contact Plate | |
Agar | |
Triple Wrapped - Irradiated |