missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Triple Wrap Sterile Pack w/VHP Indicator Tryptone Soya Agar
Carry out environmental monitoring in your most regulated environments using Thermo Scientific™ Triple Wrapped Plates, for passive and active air sampling applications.
£377.00 - £428.00
Specifications
Color | Ivory, transparent |
---|---|
Content And Storage | 2 to 25°C |
Manufacturing Origin | Germany |
Media Type | Tryptone Soya Agar |
Product Type | Agar |
Product Code | Brand | Quantity | Format | Media Type | Manufacturing Origin | Price | Quantity & Availability | ||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Format | Media Type | Manufacturing Origin | Price | Quantity & Availability | ||
15426030
|
Thermo Scientific™
PO5500B |
100/Pk. | 90mm Monoplate | Tryptone Soya Agar | Germany |
£377.00
Pack of 100 |
In Stock
Log in to see stock. |
||
15466030
|
Thermo Scientific™
PO5510D |
100/Pk. | 55mm Contact Plate | Tryptone Soya Agar | Germany |
£428.00
Pack of 100 |
In Stock
Log in to see stock. |
||
Description
Experience next-level cleanroom confidence with Thermo Scientific™ Triple Wrap Sterile Pack Tryptone Soya Agar plates, complete with VHP Indicator.
Achieve confidence in your results and ensure optimum media fertility using plates with a Vapour-phase Hydrogen Peroxide (VHP) exposure indicator, designed to confirm the integrity of the wrapping material and seal during VHP exposure.Three protective layers reduce risk of contamination:
- Primary film with moisture control patch
- Secondary film VHP barrier with immobilized desiccant sachet
- Tertiary film dust cover
Using plates with an improved shelf life and room temperature storage offers a cost-effective solution, minimizing new batch validation and refrigeration costs
Not all products are available for sale in all territories. Please inquire.
Specifications
Ivory, transparent | |
Germany | |
Agar | |
100/Pk. |
2 to 25°C | |
Tryptone Soya Agar | |
Triple Wrapped - Irradiated |
Safety and Handling
missing translation for 'shelfLife' : 43 Weeks