Learn More
Thermo Scientific™ TriPlus RSH Autosampler Fixed Needle Syringe, 100μL
Thermo Scientific™ TriPlus RSH Autosampler Fixed Needle Syringes are accurate and reproducible over a range of volumes.
Brand: Thermo Scientific™ 365H2141
Code : 16
Additional Details : Weight : 0.20000kg
Description
- Borosilicate glass with metal flanges
- Needles and plungers are stainless steel
- Field-proven syringe design
Specifications
23s G | |
Fixed Gastight Syringe | |
100 μL | |
Cone |