missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tripropargylamine, 97%, Thermo Scientific Chemicals
Click to view available options
Quantity:
1 g
5 g
25 g
Description
Tripropargylamine is used as pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Tripropargylamine is used as pharmaceutical intermediate.
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
Tripropargylamine is used as pharmaceutical intermediate.
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
Specifications
Specifications
Density | 0.927 |
Boiling Point | 79°C to 85°C (11 mmHg) |
Flash Point | 57°C (135°F) |
Linear Formula | (HC≡CCH2)3N |
Refractive Index | 1.484 |
Quantity | 25 g |
UN Number | UN1993 |
Formula Weight | 131.18 |
Percent Purity | 97% |
Chemical Name or Material | Tripropargylamine |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'