missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Triptolide, Tocris Bioscience™
Click to view available options
Quantity:
1 mg
10 mg
Description
Triptolide inhibits DNA-dependent ATPase activity of XBP and induces inhibition of RNA polymerase II (RNAPII)-mediated transcription (IC50 = 200 nM). Selective for RNAPII over RNAPI and RNAPIII. Triptolide blocks RNA synthesis in HeLa cells (IC50 = 62 nM); exhibits potent antiproliferative activity in 60 cancer cell lines (average IC50 = 12 nM) and induces apoptosis by blocking TNF-α-mediated c-IAP1 and c-IAP2 induction. Also displays immunosuppressive and anti-inflammatory activity.
Specifications
Specifications
Quantity | 10 mg |
Formula Weight | 360.4 |
Percent Purity | >98% |
Chemical Name or Material | Triptolide |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'