missing translation for 'onlineSavingsMsg'
Learn More
Triptolide, Tocris Bioscience™ Product Code.: 18045412

Triptolide, Tocris Bioscience™

Product Code. 18045412
1 mg
Click to view available options
Quantity:
1 mg
10 mg
This item is not returnable. View return policy

Product Code. 18045412

Brand: Tocris Bioscience 3253/1

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Inhibits RNAPII-mediated transcription; antitumor, anti-inflammatory and immunosuppressive

Triptolide inhibits DNA-dependent ATPase activity of XBP and induces inhibition of RNA polymerase II (RNAPII)-mediated transcription (IC50 = 200 nM). Selective for RNAPII over RNAPI and RNAPIII. Triptolide blocks RNA synthesis in HeLa cells (IC50 = 62 nM); exhibits potent antiproliferative activity in 60 cancer cell lines (average IC50 = 12 nM) and induces apoptosis by blocking TNF-α-mediated c-IAP1 and c-IAP2 induction. Also displays immunosuppressive and anti-inflammatory activity.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 38748-32-2
Molecular Formula C20H24O6
Molecular Weight (g/mol) 360.41
MDL Number MFCD00210565
InChI Key DFBIRQPKNDILPW-CIVMWXNOSA-N
Synonym triptolide, triptolid, unii-19ald1s53j, triptolide, tripterygium wilfordii, --triptolide, trisoxireno 4b,5:6,7:8a,9 phenanthro 1,2-c furan-1 3h-one, 3b,4,4a,6,6a,7a,7b,8b,9,10-decahydro-6-hydroxy-8b-methyl-6a-1-methylethyl-, 3bs,4as,5as,6r,6ar,7as,7bs,8as,8bs, triptolide, 1, triptolide pg490 /, d0i6lh, 3bs,4as,5as,6r,6ar,7as,7bs,8as,8bs-6-hydroxy-6a-isopropyl-8b-methyl-3b,4,4a,6,6a,7a,7b,8b,9,10-decahydrotrisoxireno 6,7:8a,9:4b,5 phenanthro 1,2-c furan-1 3h-one
PubChem CID 107985
ChEBI CHEBI:9747
IUPAC Name (1S,2S,4S,5S,7R,8R,9S,11S,13S)-8-hydroxy-1-methyl-7-(propan-2-yl)-3,6,10,16-tetraoxaheptacyclo[11.7.0.0²,⁴.0²,⁹.0⁵,⁷.0⁹,¹¹.0¹⁴,¹⁸]icos-14(18)-en-17-one
SMILES [H][C@@]12C[C@@H]3O[C@@]33[C@H](O)[C@]4(O[C@H]4[C@@H]4O[C@]34[C@@]1(C)CCC1=C2COC1=O)C(C)C

Specifications

Quantity 1 mg
Formula Weight 360.4
Percent Purity >98%
Chemical Name or Material Triptolide
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Triptolide, Tocris Bioscience™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.