missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals TRIS, 0.5M buffer soln., pH 6.8
Click to view available options
Quantity:
500 mL
1 L
Description
TRIS is used as a standard biological buffer in less critical applications where economy is an important consideration, component of extraction buffer e.g. phenol extraction of DNA or RNA; and as a component of separating and stacking gels in the characterization of in vitro translation products by SDS-PAGE.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
TRIS is used as a standard biological buffer in less critical applications where economy is an important consideration, component of extraction buffer e.g. phenol extraction of DNA or RNA; and as a component of separating and stacking gels in the characterization of in vitro translation products by SDS-PAGE.
Solubility
Difficult to mix.
Notes
Store in cool, dry conditions in well sealed containers. Keep container tightly closed.
TRIS is used as a standard biological buffer in less critical applications where economy is an important consideration, component of extraction buffer e.g. phenol extraction of DNA or RNA; and as a component of separating and stacking gels in the characterization of in vitro translation products by SDS-PAGE.
Solubility
Difficult to mix.
Notes
Store in cool, dry conditions in well sealed containers. Keep container tightly closed.
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Concentration | 0.5 M |
pH | 6.8 |
Physical Form | Liquid |
Quantity | 500 mL |
Solubility Information | Difficult to mix. |
Chemical Name or Material | TRIS buffer soln. |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'